Skip to Content
Merck
All Photos(6)

Key Documents

SAB2108266

Sigma-Aldrich

Anti-GAPDH Antibody

rabbit polyclonal

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
NACRES:
NA.41

Product Name

Anti-GAPDH antibody produced in rabbit, affinity isolated antibody

biological source

rabbit

Quality Level

conjugate

unconjugated

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

form

buffered aqueous solution

mol wt

36kDa

species reactivity

guinea pig, rat, horse, human, mouse

concentration

0.5 mg - 1 mg/mL

technique(s)

immunoblotting: suitable

NCBI accession no.

UniProt accession no.

shipped in

wet ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

human ... GAPDH(2597)

General description

Glyceraldehyde-3-phosphate dehydrogenase (GAPDH) localizes in the cytoplasm but can be translocated to the nucleus depending on cellular conditions. It is a tetramer containing identical chains. The gene encoding this protein is localized on human chromosome 12p13.

Immunogen

Synthetic peptide directed towards the middle region of human GAPDH

Application

Anti-GAPDH antibody produced in rabbit has been used in western blotting.

Biochem/physiol Actions

Glyceraldehyde-3-phosphate dehydrogenase (GAPDH) catalyzes the reversible oxidative phosphorylation of glyceraldehyde-phosphate, which is a critical energy-yielding step in carbohydrate metabolism. It binds to several proteins including actin, tubulin, amyloid precursor, polyglutamine peptides, DRPLA (dentatorubral-pallidoluysian atrophy) and huntingtin. Phosphorylated GAPDH associates with cytoskeletal elements and controls microtubule dynamics in the early secretory pathway. GAPDH is also a component of the functional GAIT (interferon-γ-activated inhibitor of translation) mRNP (messenger ribonucleoprotein). GAPDH expression is dysregulated during melanoma progression.

Sequence

Synthetic peptide located within the following region: KAGAHLQGGAKRVIISAPSADAPMFVMGVNHEKYDNSLKIISNASCTTNC

Physical form

Purified antibody supplied in 1x PBS buffer with 0.09% (w/v) sodium azide and 2% sucrose.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Glyceraldehyde-3-phosphate dehydrogenase is phosphorylated by protein kinase Ciota /lambda and plays a role in microtubule dynamics in the early secretory pathway.
Tisdale EJ
The Journal of Biological Chemistry, 277, 3334-3341 (2002)
Nuclear-translocated Glyceraldehyde-3-phosphate Dehydrogenase Promotes Poly(ADP-ribose) Polymerase-1 Activation during Oxidative/Nitrosative Stress in Stroke.
Nakajima H, et al.
The Journal of Biological Chemistry, 290, 14493-14503 (2015)
Mutant huntingtin: nuclear translocation and cytotoxicity mediated by GAPDH.
Bae BI, et al.
Proceedings of the National Academy of Sciences of the USA, 103, 3405-3409 (2006)
RAR?2-dependent signaling represses neuronal differentiation in mouse ES cells.
Kona SL, et al.
Differentiation, 98, 55-61 (2017)
Oxidized low-density lipoprotein decreases VEGFR2 expression in HUVECs and impairs angiogenesis.
Zhang M and Jiang L
Experimental and Therapeutic Medicine, 12, 3742-3748 (2016)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service