Skip to Content
Merck
All Photos(1)

Key Documents

SAB2102181

Sigma-Aldrich

Anti-SLC24A6 antibody produced in rabbit

affinity isolated antibody

Synonym(s):

Anti-FLJ22233, Anti-NCKX6, Anti-NCLX, Anti-Solute carrier family 24, member 6

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
NACRES:
NA.41

biological source

rabbit

Quality Level

conjugate

unconjugated

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

form

buffered aqueous solution

mol wt

64 kDa

species reactivity

human, rabbit, rat, guinea pig, bovine, mouse, dog, horse

concentration

0.5 mg - 1 mg/mL

technique(s)

western blot: suitable

UniProt accession no.

shipped in

wet ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

human ... SLC24A6(80024)

General description

Solute carrier family 24, member 6 (SLC24A6) or solute carrier family 8 (sodium/lithium/calcium exchanger), member B1 (SLC8B1) gene codes for Na+/Ca2+/Li+ exchanger (NCLX). NCLX belongs to the Na+/Ca2+ exchanger (NCX) family. This protein is expressed at a high level in the brain, skeletal and heart muscle, pancreas β-cells, and lymphocyte B-cells. NCLX contains a small regulatory domain that has a phosphorylation site and lacks Ca2+ binding domains (CBDs).

Immunogen

Synthetic peptide directed towards the middle region of human SLC24A6

Application

Anti-SLC24A6 antibody produced in rabbit has been used in western blotting(1:250).

Biochem/physiol Actions

Solute carrier family 8, member B1 (SLC8B1)/Na+/Ca2+/Li+ exchanger (NCLX) helps in the transportation of the sodium or lithium-ion in exchange for the calcium ion.

Sequence

Synthetic peptide located within the following region: SIGDAFSDFTLARQGYPRMAFSACFGGIIFNILVGVGLGCLLQISRSHTE

Physical form

Purified antibody supplied in 1x PBS buffer with 0.09% (w/v) sodium azide and 2% sucrose.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Marko Kostic et al.
Seminars in cell & developmental biology, 94, 59-65 (2019-01-19)
Mitochondrial Ca2+ transient is the earliest discovered organellar Ca2+ signaling pathway. It consist of a Ca2+ influx, mediated by mitochondrial Ca2+ uniporter (MCU), and mitochondrial Ca2+ efflux mediated by a Na+/Ca2+ exchanger (NCLX). Mitochondrial Ca2+ signaling machinery plays a fundamental
Vidhya Tangeda et al.
Cell death & disease, 13(3), 241-241 (2022-03-18)
Mitochondria are the major organelles in sensing cellular stress and inducing the response for cell survival. Mitochondrial Lon has been identified as an important stress protein involved in regulating proliferation, metastasis, and apoptosis in cancer cells. However, the mechanism of
Daniel Khananshvili
Molecular aspects of medicine, 34(2-3), 220-235 (2013-03-20)
The SLC8 gene family encoding Na(+)/Ca(2+) exchangers (NCX) belongs to the CaCA (Ca(2+)/Cation Antiporter) superfamily. Three mammalian genes (SLC8A1, SLC8A2, and SLC8A3) and their splice variants are expressed in a tissue-specific manner to mediate Ca(2+)-fluxes across the cell-membrane and thus
Marko Kostic et al.
Cell reports, 25(12), 3465-3475 (2018-12-20)
Calcium is a key regulator of mitochondrial function under both normal and pathological conditions. The mechanisms linking metabolic activity to mitochondrial Ca2+ signaling remain elusive, however. Here, by monitoring mitochondrial Ca2+ transients while manipulating mitochondrial membrane potential (ΔΨm), we found
Maria Calvo-Rodriguez et al.
Nature communications, 11(1), 2146-2146 (2020-05-03)
Mitochondria contribute to shape intraneuronal Ca2+ signals. Excessive Ca2+ taken up by mitochondria could lead to cell death. Amyloid beta (Aβ) causes cytosolic Ca2+ overload, but the effects of Aβ on mitochondrial Ca2+ levels in Alzheimer's disease (AD) remain unclear.

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service