Skip to Content
Merck
All Photos(1)

Key Documents

SAB2101148

Sigma-Aldrich

Anti-IL15 antibody produced in rabbit

affinity isolated antibody

Synonym(s):

Anti-IL-15, Anti-Interleukin 15, Anti-MGC9721

Sign Into View Organizational & Contract Pricing


About This Item

MDL number:
UNSPSC Code:
12352203
NACRES:
NA.41

biological source

rabbit

Quality Level

conjugate

unconjugated

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

form

buffered aqueous solution

mol wt

13 kDa

species reactivity

dog, horse, bovine, mouse, pig, human, sheep, rat

concentration

0.5 mg - 1 mg/mL

technique(s)

western blot: suitable

UniProt accession no.

shipped in

wet ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

human ... IL15(3600)

General description

Interleukin-15 (IL-15), a cytokine, belongs to the 4α-helix bundle cytokine family. IL-15 gene is located on human chromosome 4q31.2.

Immunogen

Synthetic peptide directed towards the N terminal region of human IL15

Biochem/physiol Actions

Interleukin-15 (IL-15) can activate inflammatory cell recruitment, angiogenesis, and synthesis of other inflammatory cytokines. It plays a key role in the progression, homeostatic proliferation, and activation of natural killer (NK) cells, natural killer T (NKT) cells, and intestinal intraepithelial lymphocytes (IELs). IL-15 plays a crucial role in the homeostatic modulation of the cluster of differentiation 8 (CD8) memory T cells. IL-15 upregulates the expression of telomerase and aids in enhancing the proliferative capacity of NK, NKT-like, and CD8 T Cells. Mutation in the IL-15 gene results in psoriasis vulgaris.

Sequence

Synthetic peptide located within the following region: RISKPHLRSISIQCYLCLLLNSHFLTEAGIHVFILGCFSAGLPKTEANWV

Physical form

Purified antibody supplied in 1x PBS buffer with 0.09% (w/v) sodium azide and 2% sucrose.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Fiona Watkinson et al.
Frontiers in immunology, 11, 594620-594620 (2021-02-05)
Interleukin-15 (IL-15) is a cytokine that has been shown to expand CD8 T cell and natural killer (NK) cell populations, and therefore has potential for potentiating adoptive immune cell therapy for cancer. Previously, IL-15 has been shown to induce proliferation
Xue-Jun Zhang et al.
The Journal of investigative dermatology, 127(11), 2544-2551 (2007-06-08)
Through a series of linkage analyses in a large Chinese family cohort of psoriasis, we previously identified and confirmed a non-HLA psoriasis linkage locus PSORS9 within a small region at 4q31.2-32.1. Within the critical region of the PSORS9 locus, IL-15
X Tan et al.
Genes and immunity, 7(5), 407-416 (2006-06-23)
Previous studies have identified mRNA three isoforms encoding interleukin-15 (IL-15) that are produced through differential splicing and encode for the same mature IL-15 protein with two different signal peptides. Our analysis of mouse intestinal epithelial cells revealed two new IL-15

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service