Skip to Content
Merck
All Photos(2)

Key Documents

SAB1408563

Sigma-Aldrich

Anti-CTBS antibody produced in rabbit

purified immunoglobulin, buffered aqueous solution

Synonym(s):

CTB

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
NACRES:
NA.41

biological source

rabbit

conjugate

unconjugated

antibody form

purified immunoglobulin

antibody product type

primary antibodies

clone

polyclonal

form

buffered aqueous solution

mol wt

antigen ~43.8 kDa

species reactivity

human

technique(s)

western blot: 1 μg/mL

NCBI accession no.

UniProt accession no.

shipped in

dry ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

human ... CTBS(1486)

General description

The gene CTBS (chitobiase) is mapped to human chromosome 1p22. The gene spans a length of approximately 20kb with seven exons interspaced by six introns. The 5′-flanking region of human chitobiase gene is rich in GC-content.

Immunogen

CTBS (NP_004379.1, 1 a.a. ~ 385 a.a) full-length human protein.

Sequence
MSRPQLRRWRLVSSPPSGVPGLALLALLALLALRLAAGTDCPCPEPELCRPIRHHPDFEVFVFDVGQKTWKSYDWSQITTVATFGKYDSELMCYAHSKGARVVLKGDVSLKDIIDPAFRASWIAQKLNLAKTQYMDGINIDIEQEVNCLSPEYDALTALVKETTDSFHREIEGSQVTFDVAWSPKNIDRRCYNYTGIADACDFLFVMSYDEQSQIWSECIAAANAPYNQTLTGYNDYIKMSINPKKLVMGVPWYGYDYTCLNLSEDHVCTIAKVPFRGAPCSDAAGRQVPYKTIMKQINSSISGNLWDKDQRAPYYNYKDPAGHFHQVWYDNPQSISLKATYIQNYRLRGIGMWNANCLDYSGDAVAKQQTEEMWEVLKPKLLQR

Biochem/physiol Actions

The gene CTBS (chitobiase) encodes a lysosomal glycosidase that functions in the degradation of asparagine-linked glycoproteins. It cleaves the reducing-end N-Acetylglucosamine residues from the chitobiose core of oligosaccharides.

Physical form

Solution in phosphate buffered saline, pH 7.4

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Lysosomal chitobiase (CTB) and the G-protein gamma 5 subunit (GNG5) genes co-localize to human chromosome 1p22.
Ahmad W
Cytogenetics and Cell Genetics, 71, 44-46 (1995)
Structure of the human gene for lysosomal di-N-acetylchitobiase.
Liu B
Glycobiology, 9, 589-593 (1999)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service