Skip to Content
Merck
All Photos(2)

Key Documents

SAB1403063

Sigma-Aldrich

Monoclonal Anti-KLF2 antibody produced in mouse

clone 1D1, purified immunoglobulin, buffered aqueous solution

Synonym(s):

LKLF

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
NACRES:
NA.41

biological source

mouse

Quality Level

conjugate

unconjugated

antibody form

purified immunoglobulin

antibody product type

primary antibodies

clone

1D1, monoclonal

form

buffered aqueous solution

mol wt

antigen ~35.79 kDa

species reactivity

human

technique(s)

indirect ELISA: suitable
western blot: 1-5 μg/mL

isotype

IgG2aκ

NCBI accession no.

UniProt accession no.

shipped in

dry ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

human ... KLF2(10365)

Related Categories

General description

Kruppel like factor 2 (KLF2) is a tumor-suppressor gene, localized on human chromosome 19p13.11.

Immunogen

KLF2 (NP_057354, 263 a.a. ~ 350 a.a) partial recombinant protein with GST tag. MW of the GST tag alone is 26 KDa.

Sequence
SWPRKRTATHTCSYAGCGKTYTKSSHLKAHLRTHTGEKPYHCNWDGCGWKFARSDELTRHYRKHTGHRPFQCHLCDRAFSRSDHLALH

Biochem/physiol Actions

Kruppel like factor 2 (KLF2) is crucial for lung functioning, cell differentiation, migration and tissue development. It modulates endothelial pro-inflammatory activation. The protein has roles in cardiovascular development and T-cell differentiation. Mutation in the gene encoding it has been associated with heritable pulmonary arterial hypertension.

Physical form

Solution in phosphate buffered saline, pH 7.4

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Kruppel-like factor 2 suppresses growth and invasion of gastric cancer cells in vitro and in vivo.
Mao QQ
Journal of Biological Regulators and Homeostatic Agents (2016)
Rafal Bartoszewski et al.
European journal of cell biology, 96(8), 758-766 (2017-10-19)
The role of microRNAs in controlling angiogenesis is recognized as a promising therapeutic target in both cancer and cardiovascular disorders. However, understanding a miRNA's pleiotropic effects on angiogenesis is a limiting factor for these types of therapeutic approaches. Using genome-wide
First identification of Kruppel-like factor 2 mutation in heritable pulmonary arterial hypertension.
Eichstaedt CA
Clinical Science (2017)
A study of the relationships between KLF2polymorphisms and body weight control in a French population
Aline Meirhaeghe
BMC Medical Genetics (2006)
W P Ries et al.
Annals of the Royal College of Surgeons of England, 101(8), 609-616 (2019-09-12)
Hypothermic machine perfusion, an organ preservation modality, involves flow of chilled preservation fluid through an allograft's vasculature. This study describes a simple, reproducible, human model that allows for interrogation of flow effects during ex vivo organ perfusion. Gonadal veins from

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service