Skip to Content
Merck
All Photos(3)

Key Documents

SAB1402157

Sigma-Aldrich

Monoclonal Anti-CRY2 antibody produced in mouse

clone 3H4, purified immunoglobulin, buffered aqueous solution

Synonym(s):

FLJ10332, HCRY2, KIAA0658, PHLL2

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
NACRES:
NA.43

biological source

mouse

Quality Level

conjugate

unconjugated

antibody form

purified immunoglobulin

antibody product type

primary antibodies

clone

3H4, monoclonal

form

buffered aqueous solution

mol wt

antigen ~36.12 kDa

species reactivity

human

technique(s)

capture ELISA: suitable
immunofluorescence: suitable
indirect ELISA: suitable
western blot: 1-5 μg/mL

isotype

IgG2aκ

NCBI accession no.

UniProt accession no.

shipped in

dry ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

human ... CRY2(1408)

General description

Cryptochrome circadian regulator 2 (CRY2) is a core circadian gene and transcriptional repressor. CRY2 gene is located on the human chromosome 11p11.2.

Immunogen

CRY2 (NP_066940.1, 141 a.a. ~ 231 a.a) partial recombinant protein with GST tag. MW of the GST tag alone is 26 KDa.

Sequence
VEVVTENSHTLYDLDRIIELNGQKPPLTYKRFQAIISRMELPKKPVGLVTSQQMESCRAEIQENHDETYGVPSLEELGFPTEGLGPAVWQG

Biochem/physiol Actions

Cryptochrome circadian regulator 2 (CRY2) is involved in DNA damage checkpoint control and cell cycle progression. It maintains the circadian rhythm through a circadian feedback loop. It is also important in immune system coordination and hematologic system development. CRY2 is a potential circadian biomarker for non-Hodgkin′s lymphoma (NHL). CRY2 is linked to rapid cycling in bipolar disorder. CRY2 functions as light-sensitive magnetosensor.

Physical form

Solution in phosphate buffered saline, pH 7.4

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Clock-Cancer Connection in Non-Hodgkin's Lymphoma: A Genetic Association Study and Pathway Analysis of the Circadian Gene Cryptochrome 2
Hoffman AE, et al.
Cancer Research, 69(8), 3605-3613 (2009)
Variants in circadian genes and prostate cancer risk: a population-based study in China
Chu Lw, et al.
Prostate Cancer and Prostatic Diseases, 11(4), 342-342 (2008)
CRY2 is associated with rapid cycling in bipolar disorder patients
Sjoholm LK, et al.
Testing, 5(9), e12632-e12632 (2010)
Human cryptochrome exhibits light-dependent magnetosensitivity
Foley LE, et al.
Nature Communications, 2, 356-356 (2011)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service