Skip to Content
Merck
All Photos(4)

Key Documents

SAB1401873

Sigma-Aldrich

Anti-INHBE antibody produced in rabbit

purified immunoglobulin, buffered aqueous solution

Synonym(s):

MGC4638

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
NACRES:
NA.43

biological source

rabbit

Quality Level

conjugate

unconjugated

antibody form

purified immunoglobulin

antibody product type

primary antibodies

clone

polyclonal

form

buffered aqueous solution

species reactivity

mouse, human

technique(s)

immunoprecipitation (IP): suitable
western blot: 1 μg/mL

NCBI accession no.

UniProt accession no.

shipped in

dry ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

human ... INHBE(83729)

General description

Inhibin subunit beta E (INHBE) is a dimeric protein, which is majorly expressed in human liver. The gene is located on human chromosome 12q13.3.

Immunogen

INHBE (NP_113667.1, 1 a.a. ~ 350 a.a) full-length human protein.

Sequence
MRLPDVQLWLVLLWALVRAQGTGSVCPSCGGSKLAPQAERALVLELAKQQILDGLHLTSRPRITHPPPQAALTRALRRLQPGSVAPGNGEEVISFATVTDSTSAYSSLLTFHLSTPRSHHLYHARLWLHVLPTLPGTLCLRIFRWGPRRRRQGSRTLLAEHHITNLGWHTLTLPSSGLRGEKSGVLKLQLDCRPLEGNSTVTGQPRRLLDTAGHQQPFLELKIRANEPGAGRARRRTPTCEPATPLCCRRDHYVDFQELGWRDWILQPEGYQLNYCSGQCPPHLAGSPGIAASFHSAVFSLLKANNPWPASTSCCVPTARRPLSLLYLDHNGNVVKTDVPDMVVEACGCS

Biochem/physiol Actions

Inhibin subunit beta E (INHBE) may be associated with carcinogenesis. Differential expression of INHBE in human endometrial tissue plays an important role in endometrial maturation and blastocyst implantation. The protein is a hepatokine linked to hepatic gene expression. It is associated with insulin resistance and body mass index in humans. The protein expression in liver is stimulated by insulin. It is involved in glucose metabolism.

Features and Benefits

Evaluate our antibodies with complete peace of mind. If the antibody does not perform in your application, we will issue a full credit or replacement antibody. Learn more.

Physical form

Solution in phosphate buffered saline, pH 7.4

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Inhibin betaE (INHBE) is a possible insulin resistance-associated hepatokine identified by comprehensive gene expression analysis in human liver biopsy samples
Sugiyama M, et al.
PLoS ONE, 13(3), e0194798-e0194798 (2018)
An insight into the phylogenetic history of HOX linked gene families in vertebrates
Abbasi AA, et al.
BMC Evolutionary Biology, 7(1), 239-239 (2007)
Evidence of inhibin/activin subunit betaC and betaE synthesis in normal human endometrial tissue
Mylonas L and Br
Reproductive Biology and Endocrinology, 8(1), 143-143 (2010)
cDNA cloning and expression of human activin betaE subunit
Hashimoto O, et al.
Molecular and Cellular Endocrinology, 194(1-2), 117-122 (2002)
Florian Bergauer et al.
Journal of molecular histology, 40(5-6), 353-359 (2009-12-25)
Inhibins are dimeric glycoproteins, composed of an alpha-subunit and one of two possible beta-subunits (betaA or betaB), with substantial roles in human reproduction and in endocrine-responsive tumours. Recently a novel beta subunit named betaE was described, although it is still

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service