Skip to Content
Merck
All Photos(1)

Key Documents

SAB1401635

Sigma-Aldrich

Anti-LAMP3 antibody produced in mouse

purified immunoglobulin, buffered aqueous solution

Synonym(s):

CD208, DC-LAMP, DCLAMP, LAMP, TSC403

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
NACRES:
NA.41

biological source

mouse

Quality Level

conjugate

unconjugated

antibody form

purified immunoglobulin

antibody product type

primary antibodies

clone

polyclonal

form

buffered aqueous solution

species reactivity

human

technique(s)

western blot: 1 μg/mL

NCBI accession no.

UniProt accession no.

shipped in

dry ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

human ... LAMP3(27074)

Immunogen

LAMP3 (AAH32940.1, 1 a.a. ~ 416 a.a) full-length human protein.

Sequence
MPRQLSAAAALFASLAVILHDGSQMRAKAFPGTRDYSQPTAAATVQDIKKPVQQPAKQAPHQTLAARFMDGHITFQTAATVKIPTTTPATTKNTATTSPITYTLVTTQATPNNSHTAPPVTEVTVGPSLAPYSLPPTITPPAHTTGTSSSTVSHTTGNTTQPSNQTTLPATLSIALHKSTTGQKPVQPTHAPGTTAAAHNTTRTAAPASTVPGPTLAPQPSSVKTGIYQVLNGSRLCIKAEMGIQLIVQDKESVFSPRRYFNIDPNATQASGNCGTRKSNLLLNFQGGFVNLTFTKDEESYYISEVGAYLTVSDPETVYQGIKHAVVMFQTAVGHSFKCVSEQSLQLSAHLQVKTTDVQLQAFDFEDDHFGNVDECSSDYTIVLPVIGAIVVGLCLMGMGVYKIRLRCQSSGYQRI

Biochem/physiol Actions

LAMP3 (lysosomal associated membrane protein 3) is tumor-specific protein and induced by hypoxia. It is known to be associated with tumors, inflammation and the maturation of dendritic cells. LAMP3 is found to be associated with Salmonella infection and promotes its intracellular proliferation. Upregulation of the gene is observed in a number of cancer such as cervical, breast, and gastrointestinal cancer and in lymph node metastasis. Increased expression of the gene shows resistance to chemotherapy and radiotherapy, and, LAMP3 might be responsible for tumor metastasis. In vitro studies point out that LAMP3 promotes invasion and migration of tumor cells.

Physical form

Solution in phosphate buffered saline, pH 7.4

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

LAMP-3 (Lysosome-Associated Membrane Protein 3) Promotes the Intracellular Proliferation of Salmonella typhimurium.
Lee EJ
Molecules and Cells, 39(7), 566-572 (2016)
Vitamin D3 regulates LAMP3 expression in monocyte derived dendritic cells.
Malaguarnera L
Cellular Immunology, 311, 13-21 (2017)
LAMP3 and TP53 overexpression predicts poor outcome in laryngeal squamous cell carcinoma.
Qiu X
International Journal of Clinical and Experimental Pathology, 8(5), 5519-5527 (2015)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service