Skip to Content
Merck
All Photos(3)

Key Documents

SAB1400673

Sigma-Aldrich

Anti-NXN antibody produced in mouse

IgG fraction of antiserum, buffered aqueous solution

Synonym(s):

Anti-FLJ12614, Anti-TRG-4

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
NACRES:
NA.41

biological source

mouse

Quality Level

conjugate

unconjugated

antibody form

IgG fraction of antiserum

antibody product type

primary antibodies

clone

polyclonal

form

buffered aqueous solution

species reactivity

human

technique(s)

immunofluorescence: suitable
western blot: 1 μg/mL

UniProt accession no.

shipped in

dry ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

human ... NXN(64359)

General description

NXN (Nucleoredoxin) is a novel cell growth regulatory protein belonging to the thioredoxin (TRX) family. It has a molecular mass of 55kDa. It is expressed in several adult tissues and is predominantly localized in the nucleus. It consists of a conserved TRX domain and a catalytic region, which exerts oxidoreductase activity.

Immunogen

NXN (NP_071908, 1 a.a. ~ 435 a.a) full-length human protein.

Sequence
MSGFLEELLGEKLVTGGGEEVDVHSLGARGISLLGLYFGCSLSAPCAQLSASLAAFYGRLRGDAAAGPGPGAGAGAAAEPEPRRRLEIVFVSSDQDQRQWQDFVRDMPWLALPYKEKHRKLKLWNKYRISNIPSLIFLDATTGKVVCRNGLLVIRDDPEGLEFPWGPKPFREVIAGPLLRNNGQSLESSSLEGSHVGVYFSAHWCPPCRSLTRVLVESYRKIKEAGQNFEIIFVSADRSEESFKQYFSEMPWLAVPYTDEARRSRLNRLYGIQGIPTLIMLDPQGEVITRQGRVEVLNDEDCREFPWHPKPVLELSDSNAAQLNEGPCLVLFVDSEDDGESEAAKQLIQPIAEKIIAKYKAKEEEAPLLFFVAGEDDMTDSLRDYTNLPEAAPLLTILDMSARAKYVMDVEEITPAIVEAFVNDFLAEKLKPEPI

Biochem/physiol Actions

NXN (Nucleoredoxin) is associated with several regulatory cellular processes such as regulation of Wnt/β-catenin pathway, oxidative stress responses. It acts as a thiol reductase and reactive oxygen species regulator during thiol-mediated oxidoreduction process. NXN possesses specific binding affinity for protein phosphatase 2A (PP2A). It interacts directly to the catalytic subunit of PP2A during phosphatase activity. It has also been reported that NXN may be involved in the transcriptional regulation of NF-κβ, AP-1, and CREB activation in HEK293 cells.

Physical form

Solution in phosphate buffered saline, pH 7.4

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

H Kurooka et al.
Genomics, 39(3), 331-339 (1997-02-01)
In a yeast artificial chromosome contig close to the nude locus on mouse chromosome 11, we identified a novel gene, nucleoredoxin, that encodes a protein with similarity to the active site of thioredoxins. Nucleoredoxin is conserved between mammalian species, and
Yosuke Funato et al.
Antioxidants & redox signaling, 9(8), 1035-1057 (2007-06-15)
Thioredoxin (TRX) family proteins are involved in various biologic processes by regulating the response to oxidative stress. Nucleoredoxin (NRX), a relatively uncharacterized member of the TRX family protein, has recently been reported to regulate the Wnt/beta-catenin pathway, which itself regulates
K Hirota et al.
Biochemical and biophysical research communications, 274(1), 177-182 (2000-07-25)
Well-established mechanisms for regulation of protein activity include thiol-mediated oxidoreduction in addition to protein-protein interactions and phosphorylation. Nucleoredoxin (NRX), glutaredoxin (GRX), and thioredoxin (TRX) have been shown to act as a potent thiol reductase and reactive oxygen species regulator. They
Katarzyna Lechward et al.
FEBS letters, 580(15), 3631-3637 (2006-06-13)
A trimeric protein phosphatase 2A (PP2A(T55)) composed of the catalytic (PP2Ac), structural (PR65/A), and regulatory (PR55/B) subunits was isolated from rabbit skeletal muscle by thiophosphorylase affinity chromatography, and contained two additional proteins of 54 and 55 kDa, respectively. The 54

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service