Skip to Content
Merck
All Photos(3)

Key Documents

SAB1100322

Sigma-Aldrich

Anti-RNF4 (176-190) antibody produced in rabbit

IgG fraction of antiserum, buffered aqueous solution

Synonym(s):

Anti-RES4-26, Anti-Ring finger protein 4, Anti-SNURF

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
NACRES:
NA.41
conjugate:
unconjugated
application:
WB
clone:
polyclonal
species reactivity:
human
citations:
5
technique(s):
western blot: 1:500-1:2,000

biological source

rabbit

Quality Level

conjugate

unconjugated

antibody form

IgG fraction of antiserum

antibody product type

primary antibodies

clone

polyclonal

form

buffered aqueous solution

mol wt

antigen ~21 kDa

species reactivity

human

technique(s)

western blot: 1:500-1:2,000

UniProt accession no.

shipped in

dry ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

human ... RNF4(6047)

General description

Really interesting new gene (RING ) finger protein 4 (RNF4) gene encodes ubiquitin E3 ligase, an enzyme widely expressed in many tissues. The protein contains a RING (really interesting new gene) finger domain and around three small ubiquitin-related modifier (SUMO)-interaction motifs. RNF4 is mapped to human chromosome 4p16.3.

Immunogen

synthetic peptide corresponding to amino acids 176-190 of human RNF4

Application

Anti-RNF4 (176-190) antibody produced in rabbit has been used in western blotting analysis.

Biochem/physiol Actions

Really interesting new gene (RING ) finger protein 4 (RNF4) catalyzes the ubiquitylation of proteins associated with small ubiquitin-related modifier (SUMO). Rnf4 is involved in DNA repair, in transcription and chromatin regulation. It is also associated with the process of hematopoiesis. RNF4 is implicated in acute promyelocytic leukemia, acute myeloid leukemia, Fanconi anemia and aplastic anemia.

Target description

RNF4 (176-190), ring finger protein 4. encodes a protein that contains a RING finger motif and acts as a transcription regulator. This protein has been shown to interact with, and inhibit the activity of, TRPS1, a transcription suppressor of GATA-mediated

Physical form

Solution in 0.01 M phosphate buffered saline, pH 7.4, containing 15 mM sodium azide.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

HSP70-Hrd1 axis precludes the oncorepressor potential of N-terminal misfolded Blimp-1s in lymphoma cells
Wang WF, et al.
Nature Communications, 8(1), 363-363 (2017)
E1B-55K-mediated regulation of RNF4 SUMO-targeted ubiquitin ligase promotes human adenovirus gene expression
Muncheberg S, et al.
Journal of Virology, 92(13), e00164-e00118 (2018)
RNF4 regulates zebrafish granulopoiesis through the DNMT1-C/EBPalpha axis
Wang L, et al.
Faseb Journal, 32(9), 4930-4940 (2018)
RNF4 is a growth inhibitor expressed in germ cells but not in human testicular tumors
Pero R, et al.
The American Journal of Pathology, 159(4), 1225-1230 (2001)
Wen-Fang Wang et al.
Nature communications, 8(1), 363-363 (2017-08-27)
B lymphocyte-induced maturation protein-1 (Blimp-1) ensures B-cell differentiation into the plasma cell stage, and its instability constitutes a crucial oncogenic element in certain aggressive cases of activated B cell-like diffuse large B-cell lymphoma (ABC-DLBCL). However, the underlying degradation mechanisms and

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service