Skip to Content
Merck
All Photos(1)

Documents

S9192

Sigma-Aldrich

Ser-Leu-Ile-Gly-Lys-Val-amide

≥95% (HPLC), solid

Synonym(s):

PAR-2 (1-6) Human, PAR2 activating peptide, PAR2-AP, SLIGKV-NH2

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C28H54N8O7
CAS Number:
Molecular Weight:
614.78
MDL number:
UNSPSC Code:
12352200
PubChem Substance ID:
NACRES:
NA.32

Quality Level

Assay

≥95% (HPLC)

form

solid

UniProt accession no.

storage temp.

−20°C

SMILES string

CC[C@H](C)[C@H](NC(=O)[C@H](CC(C)C)NC(=O)[C@@H](N)CO)C(=O)NCC(=O)N[C@@H](CCCCN)C(=O)N[C@@H](C(C)C)C(N)=O

InChI

1S/C28H54N8O7/c1-7-17(6)23(36-27(42)20(12-15(2)3)34-25(40)18(30)14-37)28(43)32-13-21(38)33-19(10-8-9-11-29)26(41)35-22(16(4)5)24(31)39/h15-20,22-23,37H,7-14,29-30H2,1-6H3,(H2,31,39)(H,32,43)(H,33,38)(H,34,40)(H,35,41)(H,36,42)/t17-,18-,19-,20-,22-,23-/m0/s1

InChI key

HOWDUIVVWDUEED-WAUHAFJUSA-N

Gene Information

mouse ... Insrr(23920)

Amino Acid Sequence

Ser-Leu-Ile-Gly-Lys-Val-NH2

Biochem/physiol Actions

Proteinase-activated receptor (PAR-2) is a member of proteolytically cleaved receptors. It is activated by a synthetic peptide (SLIGKV), that is exposed after trypsin cleavage of the amino terminus. PAR-2 stimulates an increase in cytosolic Ca2+ ion concentration.
Proteinase-activated receptor-2 (PAR2)-activating peptide.

Packaging

Bottomless glass bottle. Contents are inside inserted fused cone.

Other Notes

Lyophilized from 0.1% TFA in H2O

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Kaushik Das et al.
Scientific reports, 8(1), 7357-7357 (2018-05-11)
Metastasis, the hallmark of cancer propagation is attributed by the modification of phenotypic/functional behavior of cells to break attachment and migrate to distant body parts. Cancer cell-secreted microvesicles (MVs) contribute immensely in disease propagation. These nano-vesicles, generated from plasma membrane
D Quanjun et al.
European review for medical and pharmacological sciences, 20(22), 4688-4696 (2016-12-03)
Esophageal Cancer (EC) is a common malignant tumor occurred in the digestive tract. In this study, we investigated the mechanism of Protease Activated Receptor 2 (PAR-2) on the proliferation of esophageal cancer cell. Transfected esophageal cancer (EC) cell (PAR-2shRNA EC109)
Hendrik Ungefroren et al.
Molecular pharmacology, 92(5), 519-532 (2017-08-27)
Transforming growth factor-
Abhishek Roy et al.
The Journal of biological chemistry, 292(33), 13688-13701 (2017-05-20)
Cell migration and invasion are very characteristic features of cancer cells that promote metastasis, which is one of the most common causes of mortality among cancer patients. Emerging evidence has shown that coagulation factors can directly mediate cancer-associated complications either
Induction of vascular smooth muscle cell growth by selective activation of the proteinase activated receptor-2 (PAR-2)
Bono F, et al.
Biochemical and Biophysical Research Communications, 241(3), 762-764 (1997)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service