Skip to Content
Merck
All Photos(3)

Key Documents

S5569

Sigma-Aldrich

Anti-Superoxide Dismutase (MnSOD) (KC-19) antibody produced in rabbit

enhanced validation

IgG fraction of antiserum, buffered aqueous solution

Synonym(s):

Anti-IPO-B, Anti-Indophenoloxidase B, Anti-MNSOD, Anti-Manganese Dependent SOD, Anti-Manganese Superoxide Dismutase 2, Anti-Mitochodrial SOD, Anti-Mn-SOD, Anti-SOD2, Anti-Superoxide Dismutase 2

Sign Into View Organizational & Contract Pricing


About This Item

MDL number:
UNSPSC Code:
12352203
NACRES:
NA.41

biological source

rabbit

conjugate

unconjugated

antibody form

IgG fraction of antiserum

antibody product type

primary antibodies

clone

polyclonal

form

buffered aqueous solution

mol wt

antigen ~24 kDa

species reactivity

rat, human

enhanced validation

independent
Learn more about Antibody Enhanced Validation

technique(s)

indirect immunofluorescence: 1:250-1:500 using methanol-acetone-fixed rat NRK cells
western blot: 1:1,000-1:2,000 using whole extract of rat brain and a chemiluminescent detection reagent
western blot: 1:2,000-1:4,000 using human HeLa mitochondria extract and a chemiluminescent detection reagent

UniProt accession no.

shipped in

dry ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

human ... SOD2(6648)
mouse ... Sod2(20656)
rat ... Sod2(24787)

General description

Manganese superoxide dismutase (MnSOD) protein is a homotetrameric SOD. This protein is composed of four subunits, each containing one Mn2+. MnSOD enzyme is synthesized in the cytosol and is modified for its transport into the mitochondria where it resides in the matrix.
The SOD2 (superoxide dismutase 2) gene is mapped to human chromosome 6q25.3. The encoded protein functions as a tetramer and is localized to mitochondria.

Specificity

Anti-Superoxide Dismutase (MnSOD) (KC-19) recognizes human and rat Superoxide Dismutase (MnSOD).

Immunogen

synthetic peptide corresponding to amino acid residues 202-220 of human superoxide dismutase (MnSOD), conjugated to KLH. The corresponding sequence is identical in apes and monkeys, differs by one amino acid in mouse and horse, by two amino acids in cow, and by four amino acids in rat.

Application

Anti-Superoxide Dismutase (MnSOD) (KC-19) antibody produced in rabbit may be used in immunoblotting and indirect immunofluorescence.

Biochem/physiol Actions

Manganese superoxide dismutase (MnSOD) is essential for the survival of aerobic life. This protein is inducible by tumor necrosis factor (TNF) and protects cells from TNF-mediated apoptosis. MnSOD is proposed to function as a tumor suppressor by modulating the activity of redox-sensitive transcription factors and specific signal mediators. Overexpression of the MnSOD gene protects neurons from N-methyl-D-aspartate receptor (NMDA) and nitric oxide-induced neurotoxicity. A decline in the MnSOD activity occurs in aging, progeria, cancer, asthma and transplant rejection.
Superoxide Dismutase 2 (SOD2) is an antioxidant enzyme and catalyzes the conversion of superoxide to hydrogen peroxide and molecular oxygen. The enzyme offers protection against oxidative damage induced by reactive oxygen species and also is radioprotective. The gene is associated with the metastasis and invasion process of salivary adenoid cystic carcinoma. Manganese SOD is involved in detoxification.

Physical form

Solution in phosphate buffered saline, pH 7.4, containing 15 mM sodium azide.

Storage and Stability

For continuous use, store at 2-8 °C for up to one month. For extended storage, freeze in working aliquots. Repeated freezing and thawing is not recommended. Storage in frost-free freezers is also not recommended. If slight turbidity occurs upon prolonged storage, clarify the solution by centrifugation before use. Working dilutions should be discarded if not used within 12 hours.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

WGK

nwg

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

L A Macmillan-Crow et al.
Free radical research, 34(4), 325-336 (2001-05-01)
Manganese superoxide dismutase (MnSOD) is essential for life as dramatically illustrated by the neonatal lethality of mice that are deficient in MnSOD. In addition, mice expressing only 50% of the normal compliment of MnSOD demonstrate increased susceptibility to oxidative stress
Sumitra Miriyala et al.
Biochimica et biophysica acta, 1822(5), 794-814 (2011-12-27)
Increased understanding of the role of mitochondria under physiological and pathological conditions parallels increased exploration of synthetic and natural compounds able to mimic MnSOD - endogenous mitochondrial antioxidant defense essential for the existence of virtually all aerobic organisms from bacteria
M Gonzalez-Zulueta et al.
The Journal of neuroscience : the official journal of the Society for Neuroscience, 18(6), 2040-2055 (1998-04-18)
Neuronal nitric oxide synthase (nNOS) neurons kill adjacent neurons through the action of NMDA-glutamate receptor activation, although they remain relatively resistant to the toxic effects of NMDA and NO. The molecular basis of the resistance of nNOS neurons to toxic
S K Manna et al.
The Journal of biological chemistry, 273(21), 13245-13254 (1998-05-28)
Several recently identified intracellular proteins associate with the tumor necrosis factor (TNF) receptor and activate nuclear transcription factor (NF)-kappaB, c-Jun kinase, and apoptosis. However, the mechanism is not understood. In the present report, we investigated the role of reactive oxygen
Speranza Rubattu et al.
Journal of the American Heart Association, 5(2) (2016-02-19)
The genetic basis of stroke susceptibility remains to be elucidated. STR1 quantitative trait locus (STR1/QTL) was identified on rat chromosome 1 of stroke-prone spontaneously hypertensive rat (SHRSP) upon Japanese-style stroke-permissive diet (JD), and it contributes to 20% of the stroke

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service