Skip to Content
Merck
All Photos(3)

Key Documents

S5128

Sigma-Aldrich

Stearic anhydride

≥97.0% (GC)

Synonym(s):

Octadecanoic anhydride

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
[CH3(CH2)16CO]2O
CAS Number:
Molecular Weight:
550.94
Beilstein:
445979
EC Number:
MDL number:
UNSPSC Code:
12352211
PubChem Substance ID:
NACRES:
NA.25

Assay

≥97.0% (GC)

mp

70-72 °C (lit.)

lipid type

saturated FAs

storage temp.

−20°C

SMILES string

CCCCCCCCCCCCCCCCCC(=O)OC(=O)CCCCCCCCCCCCCCCCC

InChI

1S/C36H70O3/c1-3-5-7-9-11-13-15-17-19-21-23-25-27-29-31-33-35(37)39-36(38)34-32-30-28-26-24-22-20-18-16-14-12-10-8-6-4-2/h3-34H2,1-2H3

InChI key

WVJVHUWVQNLPCR-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Application


  • Stearic acid esterified pectin: Preparation, characterization, and application in edible hydrophobic pectin/chitosan composite films.: This research presents a novel method for esterifying pectin with stearic anhydride to create hydrophobic composite films. These films exhibit enhanced barrier properties and potential applications in food packaging, offering a biodegradable alternative to conventional materials (Wang et al., 2021).

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Michele Marin da Costa et al.
Journal of biomedical optics, 23(10), 1-11 (2018-10-24)
We present the use of Raman spectroscopy for determination of functional characteristics of insect repellents and sunscreens by identifying the active ingredients of these products applied topically to the skin. Commercial formulations of insect repellents and sunscreens (SPF 15 and
Yu Winston Wang et al.
Journal of biomedical optics, 23(4), 1-8 (2018-04-17)
Surface-enhanced Raman scattering (SERS) nanoparticles (NPs) are increasingly being engineered for a variety of disease-detection and treatment applications. For example, we have previously developed a fiber-optic Raman-encoded molecular imaging (REMI) system for spectral imaging of biomarker-targeted SERS NPs topically applied

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service