Skip to Content
Merck
All Photos(1)

Documents

S4881

Sigma-Aldrich

Staphylococcal enterotoxin B from Staphylococcus aureus

Synonym(s):

Enterotoxin B, Staphylococcal, SEB

Sign Into View Organizational & Contract Pricing


About This Item

CAS Number:
MDL number:
UNSPSC Code:
12352202
NACRES:
NA.25

contains

sodium phosphate buffer as balance

Quality Level

composition

Protein, ~25% Lowry

availability

not available in USA

impurities

Staphylococcal enterotoxin A (SEA), essentially free

application(s)

metabolomics
vitamins, nutraceuticals, and natural products

storage temp.

2-8°C

Looking for similar products? Visit Product Comparison Guide

Application

Staphylococcal enterotoxin B (SEB) which binds to MHC class II molecules and specific Vβ regions of T cell receptors (TCR), resulting in the activation of both monocytes/macrophages and T lymphocytes may be used as a superantigen and immune system activator. SEB may be used as a reference material in the development of SEB detection systems.

Biochem/physiol Actions

A superantigen for T-lymphocytes. Activator of the immune system that binds to major histocompatibility complex (MHC) antigens and CD28.

Packaging

Package size based on protein content.

related product

Product No.
Description
Pricing

Pictograms

Skull and crossbones

Signal Word

Danger

Hazard Classifications

Acute Tox. 1 Dermal - Acute Tox. 1 Inhalation - Acute Tox. 1 Oral - Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

6.1A - Combustible acute toxic Cat. 1 and 2 / very toxic hazardous materials

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Bacterial superantigens
Proft, T. and Fraser, J.D.
Clinical and Experimental Immunology, 133, 299-299 (2008)
Clarifying the mechanism of superantigen toxicity.
John D Fraser
PLoS biology, 9(9), e1001145-e1001145 (2011-09-21)
Oliver G Weingart et al.
Sensors (Basel, Switzerland), 12(2), 2324-2339 (2012-03-23)
Prevalent incidents support the notion that toxins, produced by bacteria, fungi, plants or animals are increasingly responsible for food poisoning or intoxication. Owing to their high toxicity some toxins are also regarded as potential biological warfare agents. Accordingly, control, detection
Winnie Bergstedt et al.
PloS one, 5(6), e11277-e11277 (2010-07-02)
Tuberculin is still the only available skin test reagent for the diagnosis of mycobacterial infection. The product has a remarkable sensitivity, but poor specificity. Previous studies, including two human phase I clinical trials, have indicated that rdESAT-6 has a potential
Philippe O Gannon et al.
Cytotherapy, 22(12), 780-791 (2020-10-19)
Several studies report on Good Manufacturing Process (GMP)-compliant manufacturing protocols for the ex vivo expansion of tumor-infiltrating lymphocytes (TILs) for the treatment of patients with refractory melanoma and other solid malignancies. Further opportunities for improvements in terms of ergonomy and

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service