Skip to Content
Merck
All Photos(4)

Key Documents

S2500

Sigma-Aldrich

Scopoletin

≥99%

Synonym(s):

6-Methoxyumbelliferone, 6-Methylesculetin, 7-Hydroxy-6-methoxy-2H-1-benzopyran-2-one, 7-Hydroxy-6-methoxycoumarin, Buxuletin, Chrysatropic acid, Escopoletin, Esculetin-6-methyl ether, Gelseminic acid, Murrayetin

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C10H8O4
CAS Number:
Molecular Weight:
192.17
Beilstein:
156296
EC Number:
MDL number:
UNSPSC Code:
12171501
PubChem Substance ID:
NACRES:
NA.28

Assay

≥99%

mp

203-205 °C (lit.)

SMILES string

COc1cc2C=CC(=O)Oc2cc1O

InChI

1S/C10H8O4/c1-13-9-4-6-2-3-10(12)14-8(6)5-7(9)11/h2-5,11H,1H3

InChI key

RODXRVNMMDRFIK-UHFFFAOYSA-N

Gene Information

human ... ACHE(43)

Looking for similar products? Visit Product Comparison Guide

Application

  • Production of Polyphenolic Natural Products by Bract-Derived Tissue Cultures of Three Medicinal Tilia spp.: A Comparative Untargeted Metabolomics Study.: This study investigates the production of polyphenolic compounds, including scopoletin, in tissue cultures derived from the bracts of three medicinal Tilia species. The research highlights the potential of these cultures in producing valuable natural products (Szűcs et al., 2024).

Biochem/physiol Actions

Dye that can be used to detect the release of reactive oxygen species during the oxidative burst; peroxynitrite scavenger; acetylcholinesterase inhibitor.
Scopoletin significantly increases activity of lipoprotein lipase in adipocytes by promoting transcription of the LPL gene. It also partially reverses inhibition of LPL by TNF-α.

Storage Class Code

11 - Combustible Solids

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Ae Ra Kim et al.
Phytotherapy research : PTR, 18(1), 1-7 (2004-01-30)
Peroxynitrite (ONOO(-)) is one of cytotoxic species produced by the reaction between superoxide (*O(2) (-)) and nitric oxide (NO). The main aim of this study was to identify ONOO(-) scavenging constituents from herbs. Methanolic extracts derived from one hundred thirty
Shakti Deep Pachauri et al.
Journal of ethnopharmacology, 139(1), 34-41 (2011-11-24)
Noni (Morinda citrifolia L.) is widely used for different illnesses including CNS disorders. Recently Noni has been reported to prevent amyloid beta induced memory impairment in mice. However, the influence of Noni on cholinergic system has not been explored so
J T Corbett
Journal of biochemical and biophysical methods, 18(4), 297-307 (1989-06-01)
Scopoletin, 7-hydroxy-6-methoxy-2H-1-benzopyran-2-one, a naturally occurring component in cotton leaf and citrus peel is a fluorescent substrate for peroxidase which has been used by many investigators for the determination of hydrogen peroxide concentration. The technical details of these investigations are application-specific
Kamal Massoud et al.
Plant physiology, 159(1), 286-298 (2012-03-13)
Phosphite (Phi), a phloem-mobile oxyanion of phosphorous acid (H(3)PO(3)), protects plants against diseases caused by oomycetes. Its mode of action is unclear, as evidence indicates both direct antibiotic effects on pathogens as well as inhibition through enhanced plant defense responses
An-Sheng Cheng et al.
Food & function, 3(8), 886-890 (2012-06-27)
Scopoletin is an antioxidant found in certain weedy plants. It exerts anti-inflammatory, anti-allergic, and anti-diabetic activities. It remains unknown whether scopoletin regulates T helper (Th) cells, including Th 1 and Th 2 cells. We found that scopoletin significantly inhibited phorbol

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service