Skip to Content
Merck
All Photos(1)

Key Documents

S0698

Sigma-Aldrich

Anti-SmB antibody, Mouse Monoclonal

clone 12F5, purified from hybridoma cell culture

Synonym(s):

Anti-SNRPB1, Anti-Sm protein B, Anti-Sm-B, Anti-SmB/SmB, Anti-Small nuclear ribonucleoprotein associated protein B, Anti-snRNP-B

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
NACRES:
NA.41
Pricing and availability is not currently available.

biological source

mouse

Quality Level

conjugate

unconjugated

antibody form

purified from hybridoma cell culture

antibody product type

primary antibodies

clone

12F5, monoclonal

mol wt

antigen ~25 kDa

species reactivity

human, mouse, canine

technique(s)

immunocytochemistry: suitable
immunoprecipitation (IP): suitable
indirect ELISA: suitable
western blot: 0.5-1 μg/mL using HeLa nuclear cell extract

isotype

IgG1

UniProt accession no.

shipped in

dry ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

human ... SNRPB(6628)
mouse ... Snrpb(20638)

General description

Monoclonal Anti-SmB (mouse IgG1 isotype) is derived from the hybridoma 12F5-H3.D8 produced by the fusion of mouse myeloma cells and splenocytes from BALB/c mice immunized with an SmB recombinant protein.
SNRPB (small nuclear ribonucleoprotein-associated protein B) is an important constituent of the spliceosome. It codes for SmB/B′ (small nuclear ribonucleoprotein polypeptides B and B1). It is located on human chromosome 20p13. SmB′/B are upregulated in all tissues excluding postnatal brain.

Immunogen

SmB recombinant protein.

Application

  • Monoclonal Anti-SmB antibody produced in mouse has been used in:
  • western blotting
  • enzyme-linked immunosorbent assay (ELISA)
  • immunoprecipitation
  • immunocytochemistry

Biochem/physiol Actions

SNRPB (small nuclear ribonucleoprotein-associated protein B) mutations results in cerebro-costo-mandibular syndrome. Downregulation of SNRPB affects the upregulation and processing of RNA splicing machinery components. It also plays a major role in RNA stability.

Physical form

Solution in 0.01 M phosphate buffered saline, pH 7.4, containing 15 mM sodium azide.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Allison E Drew et al.
Scientific reports, 7(1), 17993-17993 (2017-12-23)
CARM1 is an arginine methyltransferase with diverse histone and non-histone substrates implicated in the regulation of cellular processes including transcriptional co-activation and RNA processing. CARM1 overexpression has been reported in multiple cancer types and has been shown to modulate oncogenic
U7 snRNP is recruited to histone pre-mRNA in a FLASH-dependent manner by two separate regions of the stem-loop binding protein
Skrajna A, et al.
RNA, 23(6), 938-951 (2017)
Smed-SmB, a member of the LSm protein superfamily, is essential for chromatoid body organization and planarian stem cell proliferation
Fernandez-TE, et al.
Development, 137(7), 1055-1065 (2010)
Concerted regulation and molecular evolution of the duplicated SNRPB'/B and SNRPN loci
Gray TA, et al.
Nucleic Acids Research, 27(23), 4577-4584 (1999)
Functional genomics analyses of RNA-binding proteins reveal the splicing regulator SNRPB as an oncogenic candidate in glioblastoma
Correa BR, et al.
Genome Biology, 17(1) (2016)

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service