Skip to Content
Merck
All Photos(3)

Key Documents

S0652

Sigma-Aldrich

Sucrose octasulfate–aluminum complex

Synonym(s):

Sucralfate

Sign Into View Organizational & Contract Pricing


About This Item

CAS Number:
EC Number:
UNSPSC Code:
12352201
PubChem Substance ID:
NACRES:
NA.25

form

powder

Quality Level

impurities

<0.5% chloride content

color

white to almost white

storage temp.

2-8°C

SMILES string

O[Al+]O.O[Al+]O.O[Al+]O.O[Al+]O.O[Al+]O.O[Al+]O.O[Al+]O.O[Al+]O.O[Al](O)O.O[Al](O)O.O[Al](O)O.O[Al](O)O.O[Al](O)O.O[Al](O)O.O[Al](O)O.O[Al](O)O.[O-]S(=O)(=O)OC[C@H]1O[C@@H](O[C@]2(COS([O-])(=O)=O)O[C@H](OS([O-])(=O)=O)[C@@H](OS([O-])(=O)=O)[C@@H]2OS([O-])(=O)=O)[C@H](OS([O-])(=O)=O)[C@@H](OS([O-])(=O)=O)[C@@H]1OS([O-])(=O)=O

InChI

1S/C11H20O35S8.16Al.40H2O/c12-47(13,14)36-1-3-4(41-49(18,19)20)5(42-50(21,22)23)6(43-51(24,25)26)9(38-3)39-11(2-37-48(15,16)17)8(45-53(30,31)32)7(44-52(27,28)29)10(40-11)46-54(33,34)35;;;;;;;;;;;;;;;;;;;;;;;;;;;;;;;;;;;;;;;;;;;;;;;;;;;;;;;;/h3-10H,1-2H2,(H,12,13,14)(H,15,16,17)(H,18,19,20)(H,21,22,23)(H,24,25,26)(H,27,28,29)(H,30,31,32)(H,33,34,35);;;;;;;;;;;;;;;;;40*1H2/q;16*+3;;;;;;;;;;;;;;;;;;;;;;;;;;;;;;;;;;;;;;;;/p-48/t3-,4-,5+,6-,7+,8+,9+,10-,11-;;;;;;;;;;;;;;;;;;;;;;;;;;;;;;;;;;;;;;;;;;;;;;;;;;;;;;;;/m1......................................................../s1

InChI key

JTZPPHUZZDKEOC-RBQAPOGLSA-A

Gene Information

human ... PGA5(5222)

Looking for similar products? Visit Product Comparison Guide

Biochem/physiol Actions

An alkaline aluminum−sucrose complex which inhibits peptic hydrolysis and raises gastric pH.

Quality

Minimum 30% sucrose octasulfate (HPLC)

Other Notes

To gain a comprehensive understanding of our extensive range of Disaccharides for your research, we encourage you to visit our Carbohydrates Category page.

Storage Class Code

11 - Combustible Solids

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Shahram Ala et al.
World journal of surgery, 37(1), 233-238 (2012-09-27)
The aim of the present study was to evaluate the efficacy of 10 % sucralfate ointment in the reduction of acute postoperative pain after open hemorrhoidectomy. A total of 48 patients (24 men and 24 women) between 20 and 70
Jan Calissendorff et al.
Endocrine research, 37(3), 154-162 (2012-05-25)
Alcohol stimulates appetite. Ghrelin, obestatin, glucagon-like peptide 1 and leptin are putative mediators. We studied whether alcohol ingestion affects serum levels of these peripheral hormones, and if gastroprotective sucralfate prevents such an effect. Ten participants were investigated on four occasions.
Andrea A Monnig et al.
Journal of veterinary emergency and critical care (San Antonio, Tex. : 2001), 21(5), 484-495 (2012-02-10)
To review and summarize the human and veterinary literature regarding stress-related mucosal disease (SRMD) pathogenesis, patient risk factors, and therapeutic options for prophylaxis and treatment. SRMD is a common sequela of critical illness in human patients. Development of SRMD results
Melinda Hawkins et al.
International journal of colorectal disease, 27(12), 1673-1677 (2012-06-21)
Management of rectal bleeding in patients with radiation proctitis presents a conundrum for practitioners. Surgeons are appropriately concerned about using conventional methods of treatment in these patients, such as cautery for bleeding areas in the rectum, rubber band ligation, or
Ricky W McCullough
Medical hypotheses, 80(3), 230-233 (2012-12-26)
Irritable bowel syndrome (IBS), non-erosive reflux disorder (NERD), and functional dyspepsia (FD) are best classified as immuno-neuronal disorders of the mucosa or functional mucosal syndromes (FMS). Each appears to be clinically reversible using high potency sucralfate (HPS). In FMS of

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service