Skip to Content
Merck
All Photos(1)

Key Documents

S0447

Sigma-Aldrich

SIS3

≥98% (HPLC), powder

Synonym(s):

(2E)-1-(6,7-Dimethoxy-3,4-dihydro-1H-isoquinolin-2-yl)-3-(1-methyl-2-phenyl-1H-pyrrolo[2,3-b]pyridin-3-yl)-propenone hydrochloride

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C28H27N3O3 · HCl
CAS Number:
Molecular Weight:
489.99
MDL number:
UNSPSC Code:
12352200
PubChem Substance ID:
NACRES:
NA.77

Quality Level

Assay

≥98% (HPLC)

form

powder

color

yellow

solubility

H2O: <2 mg/mL
DMSO: >20 mg/mL

storage temp.

2-8°C

SMILES string

Cl.COc1cc2CCN(Cc2cc1OC)C(=O)\C=C\c3c(-c4ccccc4)n(C)c5ncccc35

InChI

1S/C28H27N3O3.ClH/c1-30-27(19-8-5-4-6-9-19)22(23-10-7-14-29-28(23)30)11-12-26(32)31-15-13-20-16-24(33-2)25(34-3)17-21(20)18-31;/h4-12,14,16-17H,13,15,18H2,1-3H3;1H/b12-11+;

InChI key

CDKIEBFIMCSCBB-CALJPSDSSA-N

Application

Sugar insensitive 3 (SIS3) has been used as smad3 inhibitor in various cells.

Biochem/physiol Actions

SIS3 is a novel specific inhibitor of TGF-β1/ALK-5 phosphorylation of Smad3
Sugar insensitive 3 (SIS3) promotes drug-induced apoptosis and hinders the function of adenosine triphosphate (ATP)-binding cassette (ABC) transporter (ABCB1) and ABCG2. It resensitizes ABCB1 and ABCG2 overexpressed in cancerous cells, which contributes to chemotherapeutics.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Transforming growth factor-beta2 utilizes the canonical Smad-signaling pathway to regulate tissue transglutaminase expression in human trabecular meshwork cells
Tovar-Vidales T, et al.
Experimental Eye Research, 93(4), 442-451 (2011)
Connective tissue growth factor is a Smad2 regulated amplifier of transforming growth factor beta actions in hepatocytes?But without modulating bone morphogenetic protein 7 signaling
Gressner OA, et al.
Hepatology, 49(6), 2021-2030 (2009)
The proto-oncogene tyrosine protein kinase Src is essential for macrophage-myofibroblast transition during renal scarring
Tang PMK, et al.
Kidney International, 93(1), 173-187 (2018)
Karl David Brown et al.
Investigative ophthalmology & visual science, 58(7), 3011-3017 (2017-06-13)
Fibrotic scarring after ocular surgeries and chemical burn injuries can impede clarity of the cornea and cause vision impairment. Transforming growth factor β (TGFβ) signaling pathway is known to mediate fibrotic scarring, and NADPH oxidase-derived reactive oxygen species has been
Ming-Heng Wu et al.
Scientific reports, 7(1), 11006-11006 (2017-09-10)
Concomitant expressions of glycan-binding proteins and their bound glycans regulate many pathophysiologic processes, but this issue has not been addressed in liver fibrosis. Activation of hepatic stellate cells (HSCs) is a rate-limiting step in liver fibrosis and is an important

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service