Skip to Content
Merck

Skip To

RTN350

GenElute Mammalian Total RNA Miniprep Kit

greener alternative

sufficient for 350 purifications

GenElute™ Mammalian Total RNA Miniprep Kit

Synonym(s):

GenElute Mammalian RNA Kit, Gen Elute

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View
Pack SizeSKUAvailabilityPrice

About This Item

NACRES:
NA.52
UNSPSC Code:
41105501
Pricing and availability is not currently available.
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


usage

sufficient for 350 purifications

Quality Level

greener alternative product characteristics

Designing Safer Chemicals
Learn more about the Principles of Green Chemistry.

sustainability

Greener Alternative Product

technique(s)

RNA purification: suitable

input

mammalian tissue
mammalian cells

test parameters

: <30 min kit run time, sample volume: up to 10^7 cells or 40 mg of tissue

greener alternative category

storage temp.

15-25°C

General description

Sigma′s GenElute Mammalian Total RNA Miniprep Kit provides a simple and convenient way to isolate total RNA from mammalian cells and tissues. Protocols are provided for cells, tissues, and fibrous tissues. These protocols differ in their cell lysis and disruption conditions. Once the RNA is bound to the GenElute Binding Column, the purification procedure is the same for all starting materials. For fibrous tissues the kit must be used with proteinase K (Cat. No. P4850) to ensure effective cell disruption. The kit combines the advantages of a silica-based system with a microspin format and eliminates the need for cesium chloride gradients, alcohol precipitation, and hazardous organic compounds such as phenol and chloroform. Cells or tissues are lysed and homogenized in a buffer containing guanidine thiocyanate to ensure thorough
denaturation of macromolecules and inactivation of RNases. Addition of ethanol causes RNA to bind when the lysate is spun through a silica membrane in a microcentrifuge tube. After washing to remove contaminants, RNA is eluted in 50-100 μL of Elution Solution. Up to 150 μg of total RNA can be isolated in less than 30 minutes.

If all traces of DNA contamination must be eliminated, further treatment with DNase I is recommended. DNase I digestion can be performed while the RNA is bound to the GenElute Binding Column using the On-Column DNase I Digestion Set (DNASE10 and DNASE70). Alternatively, for more stringent removal of contaminating DNA, the final RNA preparation can be treated with Amplification Grade DNase I (AMPD1).
We are committed to bringing you Greener Alternative Products, which adhere to one or more of The 12 Principles of Greener Chemistry. This product has Inherently Safer Chemistry, compared to the standard use of phenol and chloroform to perform DNA extractions.

Application

The purified RNA is ready for reverse transcription and PCR, labeling and microarray analysis, and other common applications. Note that RNA shorter than 200 nucleotides in length, such as tRNA, 5S rRNA, and 5.8S rRNA, is not recovered efficiently under the conditions used with this kit.
GenElute Mammalian Total RNA Miniprep Kit has been used to extract total RNA from cells and tissues.

Biochem/physiol Actions

Samples are lysed and homogenized in guanidine thiocyanate and 2-mercaptoethanol to release RNA and inactivate RNases. Lysates are spun through a filtration column to remove cellular debris and shear DNA. The filtrate is then applied to a high capacity silica column to bind total RNA, followed by washing and elution. Up to 150 μg of total RNA can be recovered per prep in 100 μl of water. The purified RNA is ready for Northern blots (Fig. 1), RT-PCR (Fig. 2) and other common applications.
The GenElute Mammalian Total RNA Miniprep Kit combines silica membrane technology with a convenient spin column format for a rapid bind, wash, and elute method to prepare high quality total RNA.

Features and Benefits

  • Purifies total RNA from up to 107 cells or 40 mg of tissue per prep
  • Yields up to 150 μg of pure, concentrated total RNA per prep
  • Recover RNA from as few as 100 cells
  • Simple and efficient–12 to 18 preps in 30 minutes
  • Faster than gravity flow anion exchange methods
  • No cumbersome steps associated with resins and magnetic slurries
  • 40% more purifications per kit than the leading supplier
  • Yields up to 150 μg of pure, concentrated total RNA per prep
  • Purifies total RNA from up to 107 cells or 40 mg of tissue per prep
  • Recover RNA from as few as 100 cells

Other Notes

For additional information, please see www.sigma-aldrich.com/totalrna.
The GenElute Mammalian Total RNA Purification Kit combines silica-membrane technology with a convenient spin column format for a rapid bind, wash, and elute method to prepare high quality total RNA.

Legal Information

GenElute is a trademark of Sigma-Aldrich Co. LLC

Compare Similar Items

View Full Comparison

Show Differences

1 of 1

This Item
RTN70RTN10DMN70
usage

sufficient for 350 purifications

usage

sufficient for 70 purifications

usage

sufficient for 10 purifications

usage

sufficient for 70 purifications

technique(s)

RNA purification: suitable

technique(s)

RNA purification: suitable

technique(s)

RNA purification: suitable

technique(s)

-

input

mammalian tissue
mammalian cells

input

mammalian tissue
mammalian cells

input

mammalian cells
mammalian tissue

input

-

test parameters

: <30 min kit run time, sample volume: up to 10^7 cells or 40 mg of tissue

test parameters

: <30 min kit run time, sample volume: up to 10^7 cells or 40 mg of tissue

test parameters

: <30 min kit run time, sample volume: up to 10^7 cells or 40 mg of tissue

test parameters

-

Quality Level

200

Quality Level

200

Quality Level

200

Quality Level

200

sustainability

Greener Alternative Product

sustainability

Greener Alternative Product

sustainability

Greener Alternative Product

sustainability

-


Still not finding the right product?


Kit Components Also Available Separately

Product No.
Description
SDS & Pricing

  • 2-Mercaptoethanol, Molecular Biology, suitable for electrophoresis, suitable for cell culture, BioReagent, 99% (GC/titration) 2
    SDS

signalword

Danger

Hazard Classifications

Acute Tox. 2 Dermal - Acute Tox. 3 Inhalation - Acute Tox. 3 Oral - Aquatic Acute 1 - Aquatic Chronic 2 - Eye Dam. 1 - Repr. 2 - Skin Corr. 1C - Skin Sens. 1 - STOT RE 2 Oral

target_organs

Liver,Heart

supp_hazards

Storage Class

6.1A - Combustible acute toxic Cat. 1 and 2 / very toxic hazardous materials

flash_point_f

154.4 °F

flash_point_c

68 °C

wgk

WGK 3



Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library



Beth Malcomson et al.
Proceedings of the National Academy of Sciences of the United States of America, 113(26), E3725-E3734 (2016-06-12)
Cystic fibrosis (CF) lung disease is characterized by chronic and exaggerated inflammation in the airways. Despite recent developments to therapeutically overcome the underlying functional defect in the cystic fibrosis transmembrane conductance regulator, there is still an unmet need to also
Directed differentiation of human induced pluripotent stem cells into functional cholangiocyte-like cells
Fotios sampaziotis
Nature Protocols, 12(4), 814-827 (2017)
Jerfiz D Constanzo et al.
Developmental biology, 415(1), 98-110 (2016-05-08)
The protein inhibitor of activated STAT-1 (PIAS1) is one of the few known SUMO E3 ligases. PIAS1 has been implicated in several biological processes including repression of innate immunity and DNA repair. However, PIAS1 function during development and tissue differentiation



Global Trade Item Number

SKUGTIN
RTN350-1KT04061836829186

Questions

  1. What are the types of RNA been extracted? m-RNA or lncRNA as well? I know about smaller and circular RNA is not efficiently extracted, is there another size limitation for the RNA been extracted? Does the RNA extracted is limited to polyA tail one only?

    1 answer
    1. The RTN kits purify total RNA, which includes mRNA, lncRNA, and all other types of RNA present in the cell. RNA does not need to contain a poly A tail for extraction. However, RNA shorter than 200 nucleotides, such as tRNA, 5S rRNA, and 5.8S rRNA, is not efficiently recovered under the conditions used with this kit.

      Helpful?

Reviews

Active Filters

  1. 1 Ratings-only review