RES0797P-A7
Potassium sulfate
Synonym(s):
Potassium sulfate
Sign Into View Organizational & Contract Pricing
All Photos(1)
About This Item
Linear Formula:
K2SO4
CAS Number:
Molecular Weight:
174.26
EC Number:
MDL number:
UNSPSC Code:
12352302
NACRES:
NA.21
Recommended Products
biological source
synthetic
Assay
98.5-101.0% dry basis
form
powder
impurities
Heavy metals; tested, Chloride; tested
suitability
suitable for manufacturing use
SMILES string
[K+].[K+].[O-]S([O-])(=O)=O
InChI
1S/2K.H2O4S/c;;1-5(2,3)4/h;;(H2,1,2,3,4)/q2*+1;/p-2
InChI key
OTYBMLCTZGSZBG-UHFFFAOYSA-L
Looking for similar products? Visit Product Comparison Guide
Related Categories
General description
To request documentation for this product, please contact Customer Support and select ‘Product Documentation′. Please note that access to the documentation for this product requires a confidentiality disclosure agreement.
Packaging
Product is available in the following package sizes:
RES0797P-A702X: 1 kg container
RES0797P-A704X: 10 kg container
RES0797P-A705X: 25 kg container
RES0797P-A702X: 1 kg container
RES0797P-A704X: 10 kg container
RES0797P-A705X: 25 kg container
Storage Class Code
13 - Non Combustible Solids
WGK
WGK 1
Flash Point(F)
Not applicable
Flash Point(C)
Not applicable
Choose from one of the most recent versions:
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Customers Also Viewed
Senthaamarai Rogawansamy et al.
International journal of environmental research and public health, 12(6), 6319-6332 (2015-06-05)
Fungal contamination in indoor environments has been associated with adverse health effects for the inhabitants. Remediation of fungal contamination requires removal of the fungi present and modifying the indoor environment to become less favourable to growth. This may include treatment
E K Doyle et al.
Veterinary parasitology, 205(1-2), 175-185 (2014-07-17)
This research was designed to determine if divergent genetic selection for resistance to Haemonchus contortus had produced correlated changes in the metabolism of amino-nitrogen in the absence or presence of H. contortus infection. Partitioning of amino acid-nitrogen between tissues was
Dana R Gradl et al.
Journal of food protection, 78(11), 1954-1959 (2015-11-12)
The survival of Salmonella on fresh ginger root (Zingiber officinale) during drying was examined using both a laboratory oven at 51 and 60°C with two different fan settings and a small commercially available food dehydrator. The survival of Salmonella in
Xinjian Shi et al.
Nature communications, 5, 4775-4775 (2014-09-03)
Tungsten trioxide/bismuth vanadate heterojunction is one of the best pairs for solar water splitting, but its photocurrent densities are insufficient. Here we investigate the advantages of using helical nanostructures in photoelectrochemical solar water splitting. A helical tungsten trioxide array is
Solimar G Machado et al.
Journal of food science, 80(8), M1842-M1849 (2015-07-21)
The storage of fresh raw milk at low temperature does not prevent proliferation of psychrotrophic bacteria that can produce heat-resistant proteolytic enzymes contributing to the reduced shelf life of dairy products. This study aimed to identify the dominant psychrotrophic proteolytic
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service