RABHRP1
HRP-conjugated Anti-Rabbit IgG Concentrate (Item I1)
Sign Into View Organizational & Contract Pricing
All Photos(1)
About This Item
UNSPSC Code:
12352203
NACRES:
NA.32
Recommended Products
antibody product type
primary antibodies
shipped in
wet ice
storage temp.
−20°C
General description
Immunoglobulin G (IgG) belongs to the immunoglobulin family and is a widely expressed serum antibody. It consists of a gamma (γ) heavy chain in the constant (C) region. The monomeric 150 kDa structure of IgG constitutes two identical heavy chains and two identical light chains with molecular weight of 50 kDa and 25 kDa, respectively. The primary structure of this antibody also contains disulfide bonds involved in linking the two heavy chains, linking the heavy and light chains and resides inside the chains. IgG is further subdivided into four classes namely, IgG1, IgG2, IgG3, and IgG4 with different heavy chains, named γ1, γ2, γ3, and γ4, respectively.
Application
HRP-conjugated Anti-Rabbit IgG Concentrate (Item I1) has been used in Western Blotting and Immunohistochemistry.
Other Notes
This item is for RAB series CBEL ELISA kits.
Not finding the right product?
Try our Product Selector Tool.
Storage Class Code
10 - Combustible liquids
WGK
WGK 1
Flash Point(F)
Not applicable
Flash Point(C)
Not applicable
Choose from one of the most recent versions:
Certificates of Analysis (COA)
Lot/Batch Number
It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.
If you need assistance, please contact Customer Support.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Customers Also Viewed
Nikita Sajeev et al.
The New phytologist, 233(1), 251-264 (2021-10-14)
Arabidopsis thaliana seed germination is marked by extensive translational control at two critical phase transitions. The first transition refers to the start of hydration, the hydration translational shift. The second shift, the germination translational shift (GTS) is the phase between
Qun Chen et al.
Journal of experimental & clinical cancer research : CR, 40(1), 160-160 (2021-05-10)
The immunosuppressive tumour microenvironment is a critical factor in the initiation and progression of glioblastoma (GBM), which is characterized by an abundance of tumour-associated macrophages (TAMs) but a paucity of infiltrating T cells. In this research, we studied whether epithelial
Systematic analysis of mRNA expression profiles in NSCLC cell lines to screen metastasis-related genes.
Liu Y, et al.
Molecular Medicine Reports, 14(6), 5093-5103 (2016)
Zhenwei Deng et al.
Frontiers in genetics, 11, 357-357 (2020-06-09)
This study was aimed to evaluate the potential function of circ-0001946 in the progression of colorectal cancer (CRC) and the related regulatory mechanism. First, the expression levels of circRNA_0001946 and microRNA-135a-5p (miR-135a-5p) in normal and CRC tissues were measured by
Constitutive Expression of GATA4 Dramatically Increases the Cardiogenic Potential of D3 Mouse Embryonic Stem Cells.
Laemmle LL, et al.
Open Biotechnology Journal, 10, 248-248 (2016)
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service