Skip to Content
Merck
All Photos(1)

Key Documents

RAB0539

Sigma-Aldrich

Human AHSG / Alpha-2-HS-glycoprotein ELISA Kit

for serum, plasma and cell culture supernatants

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41116158
NACRES:
NA.32

species reactivity

human

packaging

kit of 96 wells (12 strips x 8 wells)

technique(s)

ELISA: suitable

input

sample type plasma
sample type serum
sample type cell culture supernatant(s)

assay range

inter-assay cv: <10%
intra-assay cv: <12%
sensitivity: 0.2 ng/mL

detection method

colorimetric

shipped in

wet ice

storage temp.

−20°C

Gene Information

human ... AHSG(197)

General description

This ELISA antibody pair detects human Alpha-2-HS-glycoprotein.

Application

For research use only. Not for use in diagnostic procedures.
Please refer to the attached General ELISA KIT Procedure (sandwich, competitive & Indirect ELISA)

Biochem/physiol Actions

α 2-HS glycoprotein (AHSG), also referred to as fetuin-A, is a major serum protein component that exerts various calcium-dependent physiological activities and can mediate growth signaling in certain tumor cells. It can also act as a natural antagonist against specific TGF-β (transforming growth factor β) and BMP (bone morphogenetic protein) signaling proteins. High levels of fetuin-A in the serum is associated with mild osteogenesis imperfecta. On the other hand, reduced levels are seen in Paget′s disease. It is predicted as a potential biomarker for the early diagnosis of hypopharyngeal squamous cell carcinoma.

Other Notes

A sample Certificate of Analysis is available for this product.
Please type the word sample in the text box provided for lot number.

Kit Components Also Available Separately

Product No.
Description
SDS

  • RABTMB3ELISA Colorimetric TMB Reagent (HRP Substrate, Item H)SDS

  • RABSTOP3ELISA Stop Solution (Item I)SDS

  • RABELADAELISA 1X Assay/Sample Diluent Buffer A (Item D1)SDS

  • RABELADBELISA 5X Assay/Sample Diluent Buffer B (Item E1)SDS

  • RABWASH420X Wash Buffer (Item B)SDS

Pictograms

Corrosion

Signal Word

Warning

Hazard Statements

Precautionary Statements

Hazard Classifications

Met. Corr. 1

Storage Class Code

8A - Combustible corrosive hazardous materials

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Chia-Ter Chao et al.
Journal of the American Heart Association, 8(2), e010805-e010805 (2019-01-17)
Background Micro RNA -125b (miR-125b) has been shown to regulate vascular calcification ( VC ), and serum miR-125b levels are a potential biomarker for estimating the risk of uremic VC status. However, it is unknown whether clinical features, including chronic
Abdullahi Mudi et al.
Nephron, 136(3), 233-242 (2017-04-14)
In children with chronic kidney disease (CKD), fetuin-A and fibroblast growth factor-23 (FGF-23) have been implicated in the mechanism and progression of several cardiac changes. This study aimed to determine the types and rates of cardiac changes in children with
Ali Erdal Gunes et al.
Undersea & hyperbaric medicine : journal of the Undersea and Hyperbaric Medical Society, Inc, 44(4), 337-343 (2017-08-08)
Hyperbaric oxygen (HBO₂) treatment results in elevated production of reactive oxygen species (ROS) that leads to cellular damage. Thymoquinone (TQ) is reported to have anti-inflammatory and antimicrobial activity and may suppress the generation of free radicals. The goal of this
Margarida Sarmento-Dias et al.
Peritoneal dialysis international : journal of the International Society for Peritoneal Dialysis, 37(4), 451-457 (2017-01-18)
Fluid overload (FO) is frequently present in peritoneal dialysis (PD) patients and is associated with markers of malnutrition, inflammation, and atherosclerosis/calcification (MIAC) syndrome. We examined the relationships in stable PD patients between phase angle (PhA) and the spectrum of uremic
Claas Jasper Steffen et al.
Archives of orthopaedic and trauma surgery, 140(10), 1359-1366 (2019-12-21)
In the elderly, osteoporotic fractures of the femur are associated with high morbidity and mortality. At the time of hospitalization and during pre-operative care identification of patients at risk for poor outcome despite an otherwise good clinical condition is challenging.

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service