RAB0530
Human APP / Amyloid Beta A4 Protein ELISA Kit
for serum, plasma, cell culture supernatants and urine
Sign Into View Organizational & Contract Pricing
All Photos(1)
About This Item
UNSPSC Code:
41116158
NACRES:
NA.32
Recommended Products
species reactivity
human
packaging
kit of 96 wells (12 strips x 8 wells)
technique(s)
ELISA: suitable
input
sample type cell culture supernatant(s)
sample type urine
sample type serum
sample type plasma
assay range
inter-assay cv: <10%
intra-assay cv: <12%
sensitivity: 0.1 ng/mL
detection method
colorimetric
shipped in
wet ice
storage temp.
−20°C
Gene Information
human ... APP(351)
General description
This ELISA antibody pair detects human Amyloid beta 1-40. It has crossreactivity with mouse Amyloid beta 1-40.
Application
For research use only. Not for use in diagnostic procedures.
Please refer to the attached General ELISA KIT Procedure (sandwich, competitive & Indirect ELISA)
Please refer to the attached General ELISA KIT Procedure (sandwich, competitive & Indirect ELISA)
Other Notes
A sample Certificate of Analysis is available for this product.
Please type the word sample in the text box provided for lot number.
Please type the word sample in the text box provided for lot number.
Signal Word
Warning
Hazard Statements
Precautionary Statements
Hazard Classifications
Met. Corr. 1
Storage Class Code
8A - Combustible corrosive hazardous materials
Flash Point(F)
Not applicable
Flash Point(C)
Not applicable
Choose from one of the most recent versions:
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Li Kiang Tan et al.
Diagnostics (Basel, Switzerland), 10(1) (2019-12-28)
Dengue virus (DENV) and Zika virus (ZIKV) are flaviviruses of public health relevance. Both viruses circulate in the same endemic settings and acute infections generally manifest similar symptoms. This highlights the importance of accurate diagnosis for clinical management and outbreak
Madhura Mukhopadhyay et al.
Journal of immunology (Baltimore, Md. : 1950), 199(10), 3437-3452 (2017-10-11)
Rare patients who spontaneously control HIV replication provide a useful model to inform HIV vaccine development. HIV controllers develop particularly efficient antiviral CD4
Jennifer Manne-Goehler et al.
AIDS and behavior, 23(8), 2072-2078 (2018-12-14)
There is increasing interest in home based testing and treatment of HIV to expand access to treatment in sub-Saharan Africa. Such programs rely on self-reported HIV history and use of antiretroviral therapy (ART). However, the accuracy of self-reported ART use
Anke Heida et al.
Clinical gastroenterology and hepatology : the official clinical practice journal of the American Gastroenterological Association, 15(11), 1742-1749 (2017-06-14)
An increasing number of physicians use repeated measurements of stool calprotectin to monitor intestinal inflammation in patients with inflammatory bowel diseases (IBDs). A lateral flow-based rapid test allows patients to measure their own stool calprotectin values at home. The test
Panagiotis D Tassis et al.
BMC veterinary research, 13(1), 91-91 (2017-04-09)
Mycoplasma hyopneumoniae (M. hyo) and Porcine Circovirus Type 2 (PCV2) are major pathogens that cause significant health problems in swine worldwide. Maternal derived immunity (MDI) has been suggested as a significant immediate defence factor for newborn piglets and may interfere
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service