Skip to Content
Merck
All Photos(2)

Key Documents

RAB0479

Sigma-Aldrich

Rat Tumor Necrosis Factor α ELISA Kit

for serum, plasma and cell culture supernatant

Synonym(s):

Cachectin, TNF-alpha, Tumor necrosis factor alpha

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41116158
NACRES:
NA.84

species reactivity

rat

packaging

kit of 96 wells (12 strips x 8 wells)

technique(s)

ELISA: suitable
capture ELISA: suitable

input

sample type serum
sample type plasma
sample type cell culture supernatant(s)

assay range

inter-assay cv: <12%
intra-assay cv: <10%
sensitivity: 25 pg/mL
standard curve range: 82.3-20000 pg/mL

detection method

colorimetric

shipped in

wet ice

storage temp.

−20°C

Gene Information

rat ... Tnf(24835)

General description

The Rat TNF-α ELISA (Enzyme-Linked Immunosorbent Assay) kit is an in vitro enzyme-linked immunosorbent assay for the quantitative measurement of Rat TNF-α in serum, plasma, cell culture supernatants and urine.

Immunogen

Recombinant Rat TNF-α

Application

For research use only. Not for use in diagnostic procedures.
Please refer to the attached General ELISA KIT Procedure (sandwich, competitive & Indirect ELISA)

Other Notes

A sample Certificate of Analysis is available for this product.
Please type the word sample in the text box provided for lot number.

Kit Components Also Available Separately

Product No.
Description
SDS

  • RABELADAELISA 1X Assay/Sample Diluent Buffer A (Item D1)SDS

  • RABELADBELISA 5X Assay/Sample Diluent Buffer B (Item E1)SDS

  • RABSTOP3ELISA Stop Solution (Item I)SDS

  • RABTMB3ELISA Colorimetric TMB Reagent (HRP Substrate, Item H)SDS

  • RABWASH420X Wash Buffer (Item B)SDS

Pictograms

Corrosion

Signal Word

Warning

Hazard Statements

Precautionary Statements

Hazard Classifications

Met. Corr. 1

Storage Class Code

8A - Combustible corrosive hazardous materials


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Anlong Qi et al.
American journal of translational research, 13(4), 2784-2793 (2021-05-22)
To explore the effect of miR-21-5p on the MAP2K3 expressions and cellular apoptosis in the lung tissues of neonatal rats with hyperoxia-induced lung injuries (HILI). Twenty Sprague-Dawley neonatal rats were assigned to the normal group, and 120 rats were used
Pegah Dejban et al.
European journal of pharmacology, 887, 173579-173579 (2020-09-21)
Gastric ulcer is a prevalent disease with various etiologies, including non-steroidal anti-inflammatory drugs (NSAIDs), stress conditions, and alcohol, resulting in an inflammatory condition in the gastric mucosa. The aim of this study was to explore the protective effects of modafinil
Tao Li et al.
International journal of clinical and experimental pathology, 7(12), 8401-8410 (2015-02-13)
Anti-inflammatory and anti-apoptotic effects of polydatin (PD) have been demonstrated in our previous studies. Recently, we have found that PD treatment can ameliorate burn-induced acute lung injury (ALI). In the present study, we hypothesized that PD may provide protective effect
Jing-Yun Zhang et al.
Medical science monitor : international medical journal of experimental and clinical research, 24, 7577-7584 (2018-10-26)
BACKGROUND Protective effects of reduced beta 2 glycoprotein I (Rb2GPI) against vascular injury of diabetes mellitus have been extensively investigated. However, the effects of Rb2GPI on liver injury in diabetic animals have not been reported. MATERIAL AND METHODS A diabetic
Ting-Yu Chao et al.
Preventive nutrition and food science, 26(1), 58-66 (2021-04-17)
Hepatic ischemia/reperfusion (IR) injury is a complication that occurs during liver surgery, whereby hepatic tissue is injured by oxygen deficiency during ischemia, then further damaged by a cascade of inflammatory and oxidative insults when blood is resupplied during reperfusion. Antrodia

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service