Skip to Content
Merck
All Photos(1)

Key Documents

RAB0423

Sigma-Aldrich

Mouse E-Selectin ELISA Kit

for serum, plasma and cell culture supernatant

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41116158
NACRES:
NA.32

species reactivity

mouse

packaging

kit of 96 wells (12 strips x 8 wells)

technique(s)

ELISA: suitable
capture ELISA: suitable

input

sample type plasma
sample type serum
sample type cell culture supernatant(s)

assay range

inter-assay cv: <12%
intra-assay cv: <10%
sensitivity: 10 pg/mL
standard curve range: 12.29-3000 pg/mL

detection method

colorimetric

shipped in

wet ice

storage temp.

−20°C

Gene Information

mouse ... Sele(20339)

General description

The Mouse E-Selectin (chemokine, interferon γ- inducible T cell α chemoattractant) ELISA (Enzyme-Linked Immunosorbent Assay) kit is an in vitro enzyme-linked immunosorbent assay for the quantitative measurement of mouse E-Selectin in serum, plasma, cell culture supernatants and urine.

Immunogen

Recombinant Mouse E-Selectin

Application

For research use only. Not for use in diagnostic procedures.
Please refer to the attached General ELISA KIT Procedure (sandwich, competitive & Indirect ELISA)

Other Notes

A sample Certificate of Analysis is available for this product.
Please type the word sample in the text box provided for lot number.

Kit Components Also Available Separately

Product No.
Description
SDS

  • RABELADAELISA 1X Assay/Sample Diluent Buffer A (Item D1)SDS

  • RABELADBELISA 5X Assay/Sample Diluent Buffer B (Item E1)SDS

  • RABSTOP3ELISA Stop Solution (Item I)SDS

  • RABTMB3ELISA Colorimetric TMB Reagent (HRP Substrate, Item H)SDS

  • RABWASH420X Wash Buffer (Item B)SDS

Pictograms

Corrosion

Signal Word

Warning

Hazard Statements

Precautionary Statements

Hazard Classifications

Met. Corr. 1

Storage Class Code

8A - Combustible corrosive hazardous materials


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Justin R Ryder et al.
Metabolic syndrome and related disorders, 12(10), 509-515 (2014-08-28)
Among adolescents, obesity may increase the risk for premature cardiovascular disease (CVD). Lifestyle interventions may prevent or delay the onset of CVD through improvements in vascular health. The purpose of this study was to examine the effects of a 12-week
N Blanch et al.
Nutrition, metabolism, and cardiovascular diseases : NMCD, 24(9), 983-989 (2014-05-31)
Increased potassium intake is related to reduced blood pressure (BP) and reduced stroke rate. The effect of increased dietary potassium on endothelial function remains unknown. The aim was to determine the effect of increased dietary potassium from fruit and vegetables
I Megias-Rangil et al.
Nutrition, metabolism, and cardiovascular diseases : NMCD, 24(9), 963-968 (2014-06-08)
Obesity is associated with increased cardiovascular risk. However, the impact of morbid obesity on vascular structure and function is not well understood. This study was designed to appraise subclinical atherosclerosis markers, including carotid intima media thickness (cIMT), endothelial function, and
Isabella Werner et al.
Inflammation research : official journal of the European Histamine Research Society ... [et al.], 63(8), 649-656 (2014-05-06)
As methylene blue (MB) has been recently proposed to preserve blood pressure in case of vasoplegic syndrome and shock, an entity directly related to systemic inflammation, we aimed to elucidate the effect of MB on the expression of adhesion-molecules in
Miriam H P van Lieshout et al.
Inflammation research : official journal of the European Histamine Research Society ... [et al.], 63(11), 927-933 (2014-08-15)
To investigate the therapeutic effect of E5564 (a clinically used TLR4 inhibitor) in murine abdominal sepsis elicited by intraperitoneal infection with a highly virulent Escherichia coli in the context of concurrent antibiotic therapy. Mice were infected with different doses (~2 × 10(4)-2 × 10(6)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service