RAB0380
Human β-NGF ELISA Kit
for serum, plasma, cell culture supernatant and urine
Synonym(s):
Beta-NGF, Beta-nerve growth factor
Sign Into View Organizational & Contract Pricing
All Photos(2)
About This Item
UNSPSC Code:
41116158
NACRES:
NA.32
Recommended Products
species reactivity
human
packaging
kit of 96 wells (12 strips x 8 wells)
technique(s)
ELISA: suitable
capture ELISA: suitable
input
sample type plasma
sample type cell culture supernatant(s)
sample type serum
detection method
colorimetric
shipped in
wet ice
storage temp.
−20°C
Gene Information
human ... NGF(4803)
Related Categories
General description
The Human β-NGF (β-nerve growth factor) ELISA (Enzyme-Linked Immunosorbent Assay) kit is an in vitro enzyme-linked immunosorbent assay for the quantitative measurement of human β-NGF in serum, plasma, cell culture supernatants and urine.
Immunogen
Recombinant Human BNGF
Application
For research use only. Not for use in diagnostic procedures.
Please refer to the attached General ELISA KIT Procedure (sandwich, competitive & Indirect ELISA)
Please refer to the attached General ELISA KIT Procedure (sandwich, competitive & Indirect ELISA)
Human β-NGF (nerve growth factor) ELISA (Enzyme linked immunosorbent assay) Kit has been used for the quantification of nerve growth factor in acellularized meningeal scaffolds (AMS) and recovered meningeal tissues (referred as CMS) , in all collected medium culture wastes. It has also been used for the quantification of nerve growth factor in lysate of decellularized meningeal scaffolds (DMS).
Biochem/physiol Actions
NGFβ (nerve growth factor β ) is a potent neurotrophic factor that plays a vital role in development and maintenance of nervous systems and is essential for the differentiation and survival of neurons. It also mediates angiogenesis and facilitates progression and development of various cancer including, oral, breast and prostate cancers. Elevated expression of NGFβ contributes to progression of human cholangiocarcinoma. It has potential as marker for the diagnosis of interstitial cystitis/painful bladder syndrome (IC/PBS) and overactive bladder (OAB) and might also aid in determining the therapy for the same. It might alter placental function, as NGF signaling in placenta is influenced by antenatal depression and selective serotonin reuptake inhibitor (SSRI) treatment.
Other Notes
A sample Certificate of Analysis is available for this product.
Please type the word sample in the text box provided for lot number.
Please type the word sample in the text box provided for lot number.
Signal Word
Warning
Hazard Statements
Precautionary Statements
Hazard Classifications
Met. Corr. 1
Storage Class Code
8B - Non-combustible corrosive hazardous materials
Flash Point(F)
Not applicable
Flash Point(C)
Not applicable
Choose from one of the most recent versions:
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Prospect of umbilical cord mesenchymal stem cell culture waste in regenerative medicine
Pawitan J, et al.
Journal of Global Pharma Technology (2009)
A membrane form of guanylate cyclase is an atrial natriuretic peptide receptor.
Chinkers M
Nature, 338(6210), 78-83 (1989)
Sandeep Kumar Vishwakarma et al.
Materials science & engineering. C, Materials for biological applications, 102, 34-44 (2019-05-31)
Spinal cord injury (SCI) is one of the most precarious conditions which have been one of the major reasons for continuous increasing mortality rate of SCI patients. Currently, there is no effective treatment modality for SCI patients posing major threat
Hong-Chen Qu et al.
PloS one, 9(9), e106321-e106321 (2014-09-03)
To examine whether urinary nerve growth factor (NGF) could serve as a biomarker for interstitial cystitis/painful bladder syndrome (IC/PBS), we conducted a comprehensive meta-analysis of 9 studies. Among the studies considered, patients with IC/PBS had higher urinary NGF and NGF/Cr
Adham R Mohamed et al.
Canadian journal of physiology and pharmacology, 92(10), 813-820 (2014-09-12)
Orexin-A has been shown to modulate pain sensation and increase appetite. Rheumatoid arthritis (RA) is characterized by joint destruction, deformity, hyperalgesia, and weight reduction. Evaluate the possible effect of orexin-A on hyperalgesic and cachectic manifestations in an adjuvant-induced arthritis (AIA)
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service