Skip to Content
Merck
All Photos(2)

Documents

RAB0373

Sigma-Aldrich

Mouse pro-MMP-9 ELISA Kit

for serum, plasma and cell culture supernatant

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41116158
NACRES:
NA.32

species reactivity

mouse

packaging

kit of 96 wells (12 strips x 8 wells)

technique(s)

ELISA: suitable
capture ELISA: suitable

input

sample type plasma
sample type cell culture supernatant(s)
sample type serum

assay range

inter-assay cv: <12%
intra-assay cv: <10%
sensitivity: 50 pg/mL
standard curve range: 74.07-18000 pg/mL

detection method

colorimetric

shipped in

wet ice

storage temp.

−20°C

Gene Information

mouse ... Mmp9(17395)

General description

The Mouse pro-MMP-9 (matrix metalloproteinase-9) ELISA (Enzyme-Linked Immunosorbent Assay) kit is an in vitro enzyme-linked immunosorbent assay for the quantitative measurement of mouse pro-MMP- 9 in serum, plasma, cell culture supernatants and urine.

Immunogen

Recombinant Mouse pro-MMP9

Application

For research use only. Not for use in diagnostic procedures.
Please refer to the attached General ELISA KIT Procedure (sandwich, competitive & Indirect ELISA)

Other Notes

A sample Certificate of Analysis is available for this product.
Please type the word sample in the text box provided for lot number.

Kit Components Also Available Separately

Product No.
Description
SDS

  • RABELADEELISA 5X Assay/Sample Diluent Buffer E (Item E2)SDS

  • RABSTOP3ELISA Stop Solution (Item I)SDS

  • RABTMB3ELISA Colorimetric TMB Reagent (HRP Substrate, Item H)SDS

  • RABWASH420X Wash Buffer (Item B)SDS

Pictograms

Corrosion

Signal Word

Warning

Hazard Statements

Precautionary Statements

Hazard Classifications

Met. Corr. 1

Storage Class Code

8B - Non-combustible corrosive hazardous materials

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Javier Milara et al.
The Journal of allergy and clinical immunology, 134(2), 314-322 (2014-03-19)
Glucocorticoid functions are markedly impaired in patients with chronic obstructive pulmonary disease (COPD). The phosphodiesterase 4 inhibitor roflumilast N-oxide (RNO) is the active metabolite of roflumilast approved as a treatment to reduce the risk of exacerbations in patients with severe
Enrico Giuliani et al.
Annals of vascular surgery, 28(7), 1671-1679 (2014-06-10)
Carotid endarterectomy (CEA) is the gold standard for treating severe carotid artery stenosis, whereas carotid artery stenting (CAS) represents an endovascular alternative. The objective of this study was to assess the potential neural damage following open or endovascular carotid surgery
Yimin Zou et al.
PloS one, 9(5), e96708-e96708 (2014-05-08)
Peribronchiolar fibrosis is an important feature of small airway remodeling (SAR) in cigarette smoke-induced COPD. The aim of this study was to investigate the role of gelatinases (MMP9, MMP2) and epithelial-mesenchymal transition (EMT) in SAR related to wood smoke (WS)
Jiaye Liu et al.
Bioengineering & translational medicine, 7(1), e10263-e10263 (2022-02-04)
Besides its well-known benefits on human health, calcitriol, the hormonally active form of vitamin D3, has been being evaluated in clinical trials as an anticancer agent. However, currently available results are contradictory and not fundamentally deciphered. To the best of
Tyler Duellman et al.
Nucleic acids research, 42(9), 5518-5531 (2014-03-15)
Microribonucleic acids (miRNAs) work with exquisite specificity and are able to distinguish a target from a non-target based on a single nucleotide mismatch in the core nucleotide domain. We questioned whether miRNA regulation of gene expression could occur in a

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service