RAB0300
Mouse IL-4 ELISA Kit
for cell and tissue lysates
Sign Into View Organizational & Contract Pricing
All Photos(2)
About This Item
UNSPSC Code:
41116158
NACRES:
NA.32
Recommended Products
species reactivity
mouse
packaging
kit of 96 wells (12 strips x 8 wells)
technique(s)
ELISA: suitable
capture ELISA: suitable
input
sample type cell lysate
sample type tissue lysate
assay range
inter-assay cv: <12%
intra-assay cv: <10%
sensitivity: 1 pg/mL
standard curve range: 0.819-200 pg/mL
detection method
colorimetric
shipped in
wet ice
storage temp.
−20°C
Gene Information
mouse ... Il4(16189)
General description
The Mouse IL-4 ELISA (Enzyme-Linked Immunosorbent Assay) kit is an in vitro enzyme-linked immunosorbent assay for the quantitative measurement of mouse IL-4 in cell lysate and tissue lysate.
Immunogen
Recombinant Mouse IL-4
Application
For research use only. Not for use in diagnostic procedures.
Mouse IL-4 ELISA kit has been used for determining the levels of IL-4 in serum sample.
Mouse IL-4 ELISA kit has been used for determining the levels of IL-4 in serum sample.
For research use only. Not for use in diagnostic procedures.
Please refer to the attached General ELISA KIT Procedure (sandwich, competitive & Indirect ELISA)
Please refer to the attached General ELISA KIT Procedure (sandwich, competitive & Indirect ELISA)
Biochem/physiol Actions
IL-4 (interleukin 4) is a pleiotropic cytokine. It regulates diverse T and B cell responses including cell proliferation, survival and gene expression. IL-4 regulates the differentiation of naive CD4+ T cells into helper Th2 (T helper) cells, characterized by their cytokine-secretion profile that includes secretion of IL-4, IL-5, IL-6, IL-10, and IL-13, which favor a humoral immune response. Another dominant function of IL-4 is the regulation of immunoglobulin class switching to the IgG1 (immunoglobulin G1) and IgE isotypes. Excessive IL-4 production by Th2 cells has been associated with elevated IgE production and allergy. IL-4 helps in long-term recovery after cerebral ischemia by controlling the activities of microglia/macrophage. In mice, absence of IL-4 is associated with a state anxiety-like behavior.
Other Notes
A sample Certificate of Analysis is available for this product.
Please type the word sample in the text box provided for lot number.
Please type the word sample in the text box provided for lot number.
Kit Components Also Available Separately
Product No.
Description
SDS
Signal Word
Warning
Hazard Statements
Precautionary Statements
Hazard Classifications
Met. Corr. 1
Storage Class Code
8B - Non-combustible corrosive hazardous materials
Flash Point(F)
Not applicable
Flash Point(C)
Not applicable
Choose from one of the most recent versions:
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Flavonoid Glycosides of Polygonum capitatum protect against inflammation associated with Helicobacter pylori infection.
Zhang Shu
PLoS ONE, 10(5) (2015)
Ile50Val variant of IL4R alpha upregulates IgE synthesis and associates with atopic asthma.
Mitsuyasu H
Nature Genetics, 19, 119-120 (1998)
Interleukin (IL)-6 directs the differentiation of IL-4-producing CD4+ T cells.
Rincon M
The Journal of Experimental Medicine, 185, 461-469 (1997)
Nasal mast cells in perennial allergic rhinitics exhibit increased expression of the Fc epsilonRI, CD40L, IL-4, and IL-13, and can induce IgE synthesis in B cells.
Pawankar R
The Journal of Clinical Investigation, 99, 1492-1492 (1997)
IL-4 Knock Out Mice Display Anxiety-Like Behavior.
Moon ML
Behavior Genetics, 45, 451-460 (2015)
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service