Skip to Content
Merck
All Photos(2)

Documents

RAB0296

Sigma-Aldrich

Human IL-31 ELISA Kit

for serum, plasma, cell culture supernatant and urine

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41116158
NACRES:
NA.32

species reactivity

human

packaging

kit of 96 wells (12 strips x 8 wells)

technique(s)

ELISA: suitable
capture ELISA: suitable

input

sample type urine
sample type serum
sample type plasma
sample type cell culture supernatant(s)

assay range

inter-assay cv: <12%
intra-assay cv: <10%
sensitivity: 6 pg/mL
standard curve range: 4.92-1200 pg/mL

detection method

colorimetric

shipped in

wet ice

storage temp.

−20°C

Gene Information

human ... IL31(386653)

General description

The Human IL-31 ELISA (Enzyme-Linked Immunosorbent Assay) kit is an in vitro enzyme-linked immunosorbent assay for the quantitative measurement of human IL-31 in serum, plasma, cell culture supernatants and urine.

Immunogen

Recombinant Human IL-31

Application

For research use only. Not for use in diagnostic procedures.
Please refer to the attached General ELISA KIT Procedure (sandwich, competitive & Indirect ELISA)

Other Notes

A sample Certificate of Analysis is available for this product.
Please type the word sample in the text box provided for lot number.

Kit Components Also Available Separately

Product No.
Description
SDS

  • RABELADBELISA 5X Assay/Sample Diluent Buffer B (Item E1)SDS

  • RABELADCELISA 1X Assay/Sample Diluent Buffer C (Item L)SDS

  • RABSTOP3ELISA Stop Solution (Item I)SDS

  • RABTMB3ELISA Colorimetric TMB Reagent (HRP Substrate, Item H)SDS

  • RABWASH420X Wash Buffer (Item B)SDS

Pictograms

Corrosion

Signal Word

Warning

Hazard Statements

Precautionary Statements

Hazard Classifications

Met. Corr. 1

Storage Class Code

8A - Combustible corrosive hazardous materials


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Min Cheol Kang et al.
International journal of molecular sciences, 20(6) (2019-03-25)
Resveratrol-enriched rice (RR) was developed using genetic engineering to combine the properties of resveratrol and rice. To evaluate the effect of RR on pruritic skin inflammation in atopic dermatitis (AD)-like skin lesions, we used dinitrochlorobenzene (DNCB)-induced NC/Nga mice and an
Denis Nchang Che et al.
Molecules (Basel, Switzerland), 24(7) (2019-04-17)
IL-31 is a recently discovered cytokine that is produced not only in T-cells but also in mast cells. It is strongly implicated to play a key role in inflammatory diseases and in the pathogenesis of itch in atopic dermatitis. Apigenin
Bodoor Yaseen et al.
Rheumatology (Oxford, England), 59(9), 2625-2636 (2020-05-05)
Cytokines released by infiltrating T cells may promote mechanisms leading to fibrosis in scleroderma. The aim of this study was to investigate the role of the Th2 cytokine IL-31, and its receptor IL-31RA, in scleroderma skin and lung fibrosis. IL-31
Junji Xu et al.
Immunity, 53(2), 371-383 (2020-07-17)
Cutaneous wound healing is associated with the unpleasant sensation of itching. Here we investigated the mechanisms underlying this type of itch, focusing on the contribution of soluble factors released during healing. We found high amounts of interleukin 31 (IL-31) in
Sikai Li et al.
European journal of pharmacology, 847, 19-25 (2019-01-20)
Chronic renal failure (CRF) is a progressive disease with severe pruritus and dry skin as the major symptoms. However, the mechanisms of CRF-induced pruritus remain unclear. In this study, 5/6 nephrectomized (NPCT) mice were used as a mouse model of

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service