Skip to Content
Merck
All Photos(2)

Documents

RAB0007

Sigma-Aldrich

Human RAGE / AGER ELISA Kit

for serum, plasma, cell culture supernatant and urine

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41116158
NACRES:
NA.32

species reactivity

human

packaging

kit of 96 wells (12 strips x 8 wells)

technique(s)

ELISA: suitable
capture ELISA: suitable

input

sample type cell culture supernatant(s)
sample type plasma
sample type serum
sample type urine

assay range

inter-assay cv: <12%
intra-assay cv: <10%
sensitivity: 3 pg/mL
standard curve range: 2.06-1500 pg/mL

detection method

colorimetric

shipped in

wet ice

storage temp.

−20°C

Gene Information

human ... AGER(177)

General description

The Human RAGE ELISA (Enzyme-Linked Immunosorbent Assay) kit is an in vitro enzyme-linked immunosorbent assay for the quantitative measurement of human RAGE in serum, plasma, cell culture supernatants and urine.

Immunogen

Recombinant Human RAGE

Application

For research use only. Not for use in diagnostic procedures.

Human RAGE / AGER ELISA kit has been used for the quantitative measurement of RAGE in cerebrospinal fluid.
Please refer to the attached General ELISA KIT Procedure (sandwich, competitive & Indirect ELISA)

Biochem/physiol Actions

Advanced glycosylation end-product specific receptor (AGER) or RAGE is part of the immunoglobulin superfamily of cell surface molecules. The soluble form of the protein has a function in the Wnt pathway. RAGE has a role in the development of inflammatory and cellular damage, as well as cellular toxicity. It has been associated with diabetic atherosclerosis. AGER/RAGE also modulates cell proliferation, differentiation and apoptosis.(6}

Other Notes

A sample Certificate of Analysis is available for this product.
Please type the word sample in the text box provided for lot number.

Kit Components Also Available Separately

Product No.
Description
SDS

  • RABELADAELISA 1X Assay/Sample Diluent Buffer A (Item D1)SDS

  • RABELADBELISA 5X Assay/Sample Diluent Buffer B (Item E1)SDS

  • RABSTOP3ELISA Stop Solution (Item I)SDS

  • RABTMB3ELISA Colorimetric TMB Reagent (HRP Substrate, Item H)SDS

  • RABWASH420X Wash Buffer (Item B)SDS

Pictograms

Corrosion

Signal Word

Warning

Hazard Statements

Precautionary Statements

Hazard Classifications

Met. Corr. 1

Storage Class Code

8A - Combustible corrosive hazardous materials


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Increase of Soluble RAGE in Cerebrospinal Fluid following Subarachnoid Haemorrhage
Bartosz Sokol
BioMed Research International (2017)
Association of Polymorphisms of the Receptor for Advanced Glycation Endproducts Gene with Schizophrenia in a Han Chinese Population
Jiawu Fu
Biomedical Research, 6379639-6379639 (2017)
Circulating sRAGE in the diagnosis of osteolytic bone metastasis.
Galliera E
Journal of Biological Regulators and Homeostatic Agents, 30(4), 1203-1208 (2016)
Association of RAGE gene Gly82Ser polymorphism with coronary artery disease and ischemic stroke
Wen-Qi Ma
Medicine, 95(49), e5593-e5593 (2016)
An inverse association between serum soluble receptor of advanced glycation end products and hyperandrogenism and potential implication in polycystic ovary syndrome patients
Yu Liao
Reproductive Biology and Endocrinology, 15, 9-9 (2017)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service