Skip to Content
Merck
All Photos(6)

Key Documents

R5125

Sigma-Aldrich

Ribonuclease A from bovine pancreas

Type III-A, ≥85% RNase A basis (SDS-PAGE), 85-140 Kunitz units/mg protein

Synonym(s):

Pancreatic Ribonuclease, RNAsea, RNase A, Ribonucleate 3′-pyrimidinooligonucleotidohydrolase

Sign Into View Organizational & Contract Pricing


About This Item

CAS Number:
Enzyme Commission number:
EC Number:
MDL number:
UNSPSC Code:
12352204
NACRES:
NA.54

biological source

bovine pancreas

Quality Level

type

Type III-A

Assay

≥85% RNase A basis (SDS-PAGE)

form

lyophilized powder

specific activity

85-140 Kunitz units/mg protein

mol wt

~13,700

technique(s)

cell based assay: suitable

impurities

salt, essentially free

suitability

suitable for molecular biology

application(s)

diagnostic assay manufacturing

foreign activity

protease, essentially free

storage temp.

−20°C

SMILES string

[nH]1cnc(c1)CC(NC(=O)CCN)C(=O)O

InChI

1S/C9H14N4O3/c10-2-1-8(14)13-7(9(15)16)3-6-4-11-5-12-6/h4-5,7H,1-3,10H2,(H,11,12)(H,13,14)(H,15,16)

InChI key

CQOVPNPJLQNMDC-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

RNase A, Ribonuclease A, is an endoribonuclease that cleaves the phosphodiester bonds of single strand RNA after pyrimidine nucleotides. It attacks at the 3′ phosphate end (For example pG-pG-pC-pA-pG will be cleaved to give pG-pG-pCp and A-pG). The highest activity is exhibited with single stranded RNA. RNase A is a single chain polypeptide containing 4 disulfide bridges. In contrast to RNase B, it is not a glycoprotein. Ribonucleases do not hydrolyze DNA, because the DNA lacks 2′-OH groups essential for the formation of cyclic intermediates. RNase A can also hydrolyze RNA from protein samples. RNase A can be inhibited by alkylation of His12 and His119 and activated by potassium and sodium salts. RNAse is inhibited in the presence of heavy metal ions. RNase is also inhibited competitively by DNA.

Application

  • RNase A is used to remove RNA from DNA plasmid and genomic DNA preparations and protein samples.
  • RNase A is also used in RNA sequence analysis and protection assays.
  • RNase A has been used as a tool for computer-aided drug design.
  • RNase A supports the analysis of RNA sequences.
  • RNase A hydrolyze RNA contained in protein samples.
  • Purification of DNA is supported by RNase A.
Ribonuclease A is used to remove RNA from DNA plasmid preparations and protein samples. Ribonuclease A is used for RNase protection assays, to remove unspecifically bound RNA, analysis of RNA sequences, to hydrolyze RNA contained in protein samples, and the purification of DNA. Ribonuclease A from bovine pancreas has been used in a study to assess nickel-dependent oxidative cross-linking of a protein. Ribonuclease A from bovine pancreas has also been used in a study to investigate equilibrium constants for nonspecific binding of proteins to DNA.

Biochem/physiol Actions

Ribonuclease A is an endoribonuclease that cleaves single stranded RNA after pyrimidine nucleotides. It attacks at the 3′ phosphate end. Ribonucleases do not hydrolyze DNA, because the DNA lacks 2′-OH groups essential for the formation of cyclic intermediates. RNase can also hydrolyze RNA from protein samples. RNase A can be inhibited by alkylation of His12 and His119 and activated by potassium and sodium salts.

Features and Benefits

Our highly stable Ribonuclease A, RNase A, is suitable for removal of RNA, RNA sequencing, and DNA purification.

Preparation Note

Salt fractionated and chromatographically purified.

Analysis Note

Protein determined by E.

inhibitor

Product No.
Description
Pricing

Pictograms

Health hazard

Signal Word

Danger

Hazard Statements

Precautionary Statements

Hazard Classifications

Resp. Sens. 1

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

G Gill et al.
Chemical research in toxicology, 10(3), 302-309 (1997-03-01)
A model protein, ribonuclease A (bovine pancreas), was examined for its ability to coordinate Ni2+ and promote selective oxidation. In the presence of a peracid such as monopersulfate, HSO5-, nickel induced the monomeric RNase A to form dimers, trimers, tetramers
E S Jenuwine et al.
Analytical biochemistry, 242(2), 228-233 (1996-11-15)
Quantitative zonal DNA affinity chromatography may be used to determine accurate equilibrium constants for the binding of proteins nonspecifically to DNA. Zonal quantitative affinity chromatography has not previously been applied to the determination of binding constants of proteins to DNA
J Yu et al.
Cytometry, 14(4), 428-432 (1993-01-01)
DNA content by flow cytometry was assessed in 47 cases from a series of 130 patients with non-small cell lung carcinoma (NSCLC) given radiation therapy postoperatively. This was done in an attempt to identify which patients might benefit, or not
J Castro et al.
Cytometry, 14(7), 793-804 (1993-10-01)
An easy method for preparation of bare cell nuclei from fresh solid tissues for DNA flow cytometry is described. Pieces of up to 2 x 2 x 2 mm3 size from fresh tissues were fixed in formalin. After removal of
Thomas Blasi et al.
Nature communications, 7, 10256-10256 (2016-01-08)
Imaging flow cytometry combines the high-throughput capabilities of conventional flow cytometry with single-cell imaging. Here we demonstrate label-free prediction of DNA content and quantification of the mitotic cell cycle phases by applying supervised machine learning to morphological features extracted from

Protocols

This procedure may be used for determination of Ribonuclease A (RNase A) activity.

This procedure may be used for determination of Ribonuclease A (RNase A) activity.

This procedure may be used for determination of Ribonuclease A (RNase A) activity.

This procedure may be used for determination of Ribonuclease A (RNase A) activity.

Chromatograms

application for HPLC

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service