Skip to Content
Merck
All Photos(1)

Key Documents

PZ0390

Sigma-Aldrich

PF-06747711

≥98% (HPLC)

Synonym(s):

3-Cyano-N-(3-(1-isobutyrylpiperidin-4-yl)-1-methyl-4-(trifluoromethyl)-1H-pyrrolo[2,3-b]pyridin-5-yl)benzamide, 3-Cyano-N-[1-methyl-3-[1-(2-methyl-1-oxopropyl)-4-piperidinyl]-4-(trifluoromethyl)-1H-pyrrolo[2,3-b]pyridin-5-yl]benzamide

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C26H26F3N5O2
CAS Number:
Molecular Weight:
497.51
UNSPSC Code:
12352200
NACRES:
NA.77

Assay

≥98% (HPLC)

form

powder

color

white to beige

solubility

DMSO: 2 mg/mL, clear

storage temp.

2-8°C

SMILES string

N#CC1=CC(C(NC2=CN=C(N(C)C=C3C4CCN(C(C(C)C)=O)CC4)C3=C2C(F)(F)F)=O)=CC=C1

InChI key

BIHPJIJLDNUDGH-UHFFFAOYSA-N

Biochem/physiol Actions

PF-06747711 is a mono-imine compound. It regulates gene expression in signaling pathways by targeting a nuclear hormone receptor. It is also used to treat psoriasis.
PF-06747711 is a potent and selective RORC2 modulator.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Unifying Mechanism Involving Electron Affinity of Conjugated Imine as Anti-Cancer Agents and Miscellaneous Drugs: Reactive Oxygen Species, Oxidative Stress, and Electron Transfer
Kovacic P, et al.
Chronicles of Pharmaceutical Science, 5 (2020)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service