Skip to Content
Merck
All Photos(1)

Key Documents

PZ0344

Sigma-Aldrich

UK-371,804 HCl

≥98% (HPLC)

Synonym(s):

2-(((4-Chloro-1-guanidino-7-isoquinolinyl)sulfonyl)amino)-isobutyric acid hydrochloride, N-[[1-[(aminoiminomethyl)amino]-4-chloro-7-isoquinolinyl]sulfonyl]-2-methyl-alanine hydrochloride, UK-371804 HCl

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C14H16ClN5O4S · HCl
CAS Number:
Molecular Weight:
422.29
UNSPSC Code:
12352200
NACRES:
NA.77

Quality Level

Assay

≥98% (HPLC)

form

powder

color

white to beige

solubility

DMSO: 10 mg/mL, clear (warmed)

shipped in

dry ice

storage temp.

−20°C

SMILES string

CC(C)(NS(=O)(C1=CC2=C(C=C1)C(Cl)=CN=C2NC(N)=N)=O)C(O)=O.[H]Cl

InChI

1S/C14H16ClN5O4S.ClH/c1-14(2,12(21)22)20-25(23,24)7-3-4-8-9(5-7)11(19-13(16)17)18-6-10(8)15;/h3-6,20H,1-2H3,(H,21,22)(H4,16,17,18,19);1H

InChI key

IUBFETAFRGVODB-UHFFFAOYSA-N

Biochem/physiol Actions

UK-371,804 is a reversible, substrate-competitive, highly potent (Ki = 10 nM) and selective urokinase-type plasminogen activator (uPA) Inhibitor with excellent selectivity over tPA, plasmin, Factor IXa and Xa. When applied topically, UK-371,80 effectively penetrates into excisional wounds of experimental pigs with no adverse effect on wound closure. UK-371,804 effectively blocks EGR-FMK from binding human uPA even in the presence of tissue proteins.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service