Skip to Content
Merck
All Photos(4)

Key Documents

PVP1

Sigma-Aldrich

Poly(vinylpyrrolidone)–Iodine complex

Synonym(s):

PVP-Iodine, Polyvinylpyrrolidone Iodine, PVP-I, Povadyne® antiseptic

Sign Into View Organizational & Contract Pricing


About This Item

CAS Number:
MDL number:
UNSPSC Code:
51102829
PubChem Substance ID:
NACRES:
NA.85

form

powder

solubility

H2O: 100 mg/mL

antibiotic activity spectrum

Gram-negative bacteria
Gram-positive bacteria
fungi
parasites
viruses
yeast

Mode of action

protein synthesis | interferes

SMILES string

II.C=CN1CCCC1=O

InChI

1S/C6H9NO.I2/c1-2-7-5-3-4-6(7)8;1-2/h2H,1,3-5H2;

InChI key

CPKVUHPKYQGHMW-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Application

Poly(vinylpyrrolidone)–Iodine complex, or PVP-I, has been used to study microbial decontamination of human donor eyes and the inactivation of infectious hematopoietic necrosis virus.

Biochem/physiol Actions

PVP-1 is used as a slow iodine release formulation that iodinates lipids and oxidizes cell compounds such as tyrosine sidechains. It is used as a carbon nanotube coating agent to improve water solubility.

Other Notes

An iodophor; "tamed iodine."
Keep container tightly closed in a dry and well-ventilated place.

Legal Information

Povadyne is a registered trademark of National Patent Development Corp.

Pictograms

CorrosionEnvironment

Signal Word

Danger

Hazard Statements

Hazard Classifications

Aquatic Chronic 2 - Eye Dam. 1 - Skin Irrit. 2

Storage Class Code

11 - Combustible Solids

WGK

WGK 2

Flash Point(F)

>212.0 °F - Not applicable

Flash Point(C)

> 100 °C - Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Yangzi Jiang et al.
Acta biomaterialia, 9(9), 8089-8098 (2013-05-28)
Polyvinylpyrrolidone-iodine (Povidone-iodine, PVP-I) is widely used as an antiseptic agent for lavation during joint surgery; however, the biological effects of PVP-I on cells from joint tissue are unknown. This study examined the biocompatibility and biological effects of PVP-I on cells
Lina Du et al.
Wound repair and regeneration : official publication of the Wound Healing Society [and] the European Tissue Repair Society, 20(6), 904-910 (2012-11-01)
In this study, a multifunctional in situ-forming hydrogel (MISG) was prepared as a wound dressing designed to stop bleeding, inhibit inflammation, relieve pain, and improve healing. A mixture of poloxamers 407 and 188 was used for the matrix of the
Pracha Nuntnarumit et al.
Infection control and hospital epidemiology, 34(4), 430-432 (2013-03-08)
We conducted a randomized controlled trial in neonates with birth weight greater than or equal to 1,500 g that compared 1% aqueous chlorhexidine gluconate (CHG) with 10% povidone-iodine (PI) as a topical antiseptic. We found 1% CHG to be more
Philippe Seguin et al.
Critical care medicine, 42(1), 1-8 (2013-10-10)
To evaluate the efficacy and safety of oral care with povidone-iodine on the occurrence of ventilator-associated pneumonia in a high-risk population. A multicenter, placebo-controlled, randomized, double-blind, two-parallel-group trial performed between May 2008 and May 2011. Six ICUs in France. One
A Tuna et al.
Australian dental journal, 57(4), 435-439 (2012-11-29)
The aim of this pilot study was to evaluate the effects of mouthrinses containing 7.5% povidone iodine and 0.2% chlorhexidine on bacteraemia following impacted third molar surgery. Thirty-four patients were enrolled in this study. Patients in the first group were

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service