Skip to Content
Merck

PP0510

GlycoProfile IV Chemical Deglycosylation Kit

TFMS Deglycosylation System

Synonym(s):

Deglycosylation kit

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View

About This Item

NACRES:
NA.32
UNSPSC Code:
12352200
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


Quality Level

storage temp.

2-8°C

General description

The GlycoProfile IV Kit utilizes trifluoromethane sulfonic acid (TFMS) in a deglycosylation system that completely removes all N- and O-linked glycans while safeguarding the protein or polypeptide structure. TFMS hydrolysis of glycoproteins results in minimal protein degradation while the released glycans are eliminated. The conditions are optimized to deglycosylate simple or low molecular weight glycoproteins in 30 mins without the scavenger. For high molecular weight or complex, non-mammalian glycoproteins, anhydrous anisole is provided as a scavenger to give a high protein yield.

Application

GlycoProfile IV Chemical Deglycosylation Kit has been used in the deglycosylation of human α-L-iduronidase and peritrophic matrix from the gut of the caterpillar, Helicoverpa armigera. It has also been used to estimate the glycosylation of plant-derived recombinant human interleukin-13 (rhIL-13).

Features and Benefits

  • Eliminates both N- and O-linked glycans from proteins
  • This enables sequence analysis of the protein core by mass spectrometry (MS) or DNA-to-protein sequence comparison
  • Eliminates glycans which affect the molecular radius
  • Allows molecular mass determination by electrophoresis
  • Complete deglycosylation in as short as 30 mins for high throughput
  • Quick as it eliminates the need for multiple enzyme reactions
  • Accommodates a broad range of glycoproteins
  • Minimal degradation of the protein core to facilitate reliable MS results

Preparation Note

The kit contains sufficient reagents for 10 reactions with a sample size between 1-2 mg of a typical glycoprotein. An RNase B glycoprotein standard is included.

Legal Information

GlycoProfile is a trademark of Sigma-Aldrich Co. LLC


Kit Components Only

Product No.
Description

  • Trifluoromethanesulfonic Acid (TFMS) 5 x 1

  • RNase B Glycoprotein Standard 3 x 1

  • Pyridine solution, 60% 10 mL

  • Bromophenol blue solution, 0.2% .5 mL

  • Anisole, anhydrous 5 x 1

  • Reaction vials 10 ea

signalword

Danger

Hazard Classifications

Acute Tox. 3 Dermal - Acute Tox. 4 Inhalation - Acute Tox. 4 Oral - Eye Dam. 1 - Flam. Liq. 2 - Met. Corr. 1 - Resp. Sens. 1 - Skin Corr. 1B - STOT SE 1 - STOT SE 3

target_organs

Eyes,Central nervous system, Respiratory system

Storage Class

3 - Flammable liquids



Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library


Related Content

Glycobiology and Glycoproteomics Brochure


A novel platform for biologically active recombinant human interleukin-13 production
David J W, et al.
Plant Biotechnology (2008)
Proteomic analysis of the peritrophic matrix from the gut of the caterpillar, Helicoverpa armigera
Peter M C, et al.
Insect Biochemistry and Molecular Biology, 950-958 (2008)
Production and characterization of soluble human lysosomal enzyme alpha-iduronidase with high activity from culture media of transgenic tobacco BY-2 cells
Fu L H, et al.
Plant Science, 177(6), 668-675 (2009)



Global Trade Item Number

SKUGTIN
PP0510-1KT04061826739792