Skip to Content
Merck

PLA0079

Rabbit anti-PCNA Antibody, Affinity Purified

Powered by Bethyl Laboratories, Inc.

Synonym(s):

ATLD2, DNA polymerase delta auxiliary protein, PCNA, cyclin

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View

About This Item

NACRES:
NA.41
UNSPSC Code:
12352203
Conjugate:
unconjugated
Clone:
polyclonal
Application:
FACS, IHC, IP, WB
Citations:
7
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


biological source

rabbit

Quality Segment

conjugate

unconjugated

antibody form

affinity purified immunoglobulin

antibody product type

primary antibodies

clone

polyclonal

species reactivity

human, mouse

technique(s)

flow cytometry: 0.03 μg, immunohistochemistry: 1:2,000- 1:10,000, immunoprecipitation (IP): 2- 5 μg/mg, western blot: 1:1,000- 1:10,000

accession no.

P12004

UniProt accession no.

shipped in

wet ice

storage temp.

2-8°C

target post-translational modification

unmodified

Gene Information

rabbit ... PCNA(5111)

General description

Proliferating cell nuclear antigen (PCNA), a single-copy gene, spans about 12 kb. This gene is located on human chromosome 20p13. PCNA is expressed at high levels in the thymus, bone marrow, fetal liver, and some cells of the small intestine and colon. It is located in the nucleus.

Immunogen

The epitope recognized by PLA0079 maps to a region between residues 75 and 125 of human proliferating cell nuclear antigen using the numbering given in SwissProt entry P12004 (GeneID 5111).

Application

Rabbit anti-PCNA Antibody, Affinity Purified has been used in:
  • proximity ligation assay (PLA) (1:500)
  • immunoblotting (1:2000)
  • flow cytometry analysis

Biochem/physiol Actions

Proliferating cell nuclear antigen (PCNA) protein functions as an auxiliary factor of polymerase δ. It acts as a cofactor to polymerase ε, polymerase β, and translesion synthesis polymerases, such as η, κ, λ, and θ. In chronic lymphoid leukemia (CLL) patients, intracellular levels of PCNA protein can be used as a marker to identify clinical behavior and survival.

Physical form

Tris-citrate/phosphate buffer, pH 7 to 8 containing 0.09% Sodium Azide

Other Notes

PCNA (Proliferating Cell Nuclear Antigen) was first identified as a factor expressed in the nuclei of replicating cells. PCNA plays a role in both DNA synthesis and repair. PCNA functions as a processivity factor for DNA polymerase by acting as a DNA clamp that tethers the polymerase catalytic units to the DNA template. The structure of PCNA is that of a trimeric ring that encircles and slides along replicating DNA. PCNA interacts with and is regulated by several proteins involved in cell cycle control and check point processes.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.


Still not finding the right product?

or

Try our Product Selector Tool to narrow your options


Storage Class

12 - Non Combustible Liquids

wgk

nwg

flash_point_f

Not applicable

flash_point_c

Not applicable



Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library