Skip to Content
Merck
All Photos(1)

Key Documents

P6838

Millipore

Peptone from milk solids

Refined hydrolysate

Synonym(s):

Peptone from animal tissue, PNM, Peptonized milk nutrient

Sign Into View Organizational & Contract Pricing


About This Item

CAS Number:
MDL number:
UNSPSC Code:
41106212
NACRES:
NA.85

biological source

bovine milk

form

powder

nitrogen analysis

1.7% amino, 5.4% total

application(s)

microbiology

SMILES string

O(C)C(=O)C(CCC(CC)(CC)C)C(=O)OC

InChI

1S/C13H24O4/c1-6-13(3,7-2)9-8-10(11(14)16-4)12(15)17-5/h10H,6-9H2,1-5H3

InChI key

AIUDWMLXCFRVDR-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Seok Hoon Hong et al.
Nature communications, 3, 613-613 (2012-01-05)
To utilize biofilms for chemical transformations in biorefineries they need to be controlled and replaced. Previously, we engineered the global regulator Hha and cyclic diguanylate-binding BdcA to create proteins that enable biofilm dispersal. Here we report a biofilm circuit that
V I Iliukhin et al.
Antibiotiki i khimioterapiia = Antibiotics and chemoterapy [sic], 52(1-2), 18-20 (2008-05-09)
Principles and procedure for rapid estimation of bacteria susceptibility to antibiotics on a glucose-tryptone medium with an indicator are described. The results of the tests with 50 microbial strains of 17 species showed practically complete identity to the results of
H B Nielsen et al.
Environmental technology, 28(8), 905-914 (2007-09-21)
Two thermophilic continuously stirred tank reactors, R1 and R2, were subject to pulses of tryptone and ammonia. R1 was operated at an ammonia-N concentration of 3.0 g l(-1) and R2 was operated at an ammonia-N concentration of 1.7 g l(-1).
Priyadharshini Ramachandran et al.
Journal of microbiology and biotechnology, 23(3), 351-356 (2013-03-07)
A high beta-glucosidase (BGL)-producing strain, Stereum hirsutum, was identified and isolated and showed a maximum BGL activity (10.4 U/ml) when cultured with Avicel and tryptone as the carbon and nitrogen sources, respectively. In comparison with other BGLs, BGL obtained from
Takashi Kuda et al.
Food chemistry, 134(4), 1719-1723 (2013-02-28)
Listeria monocytogenes causes listeriosis in humans, mainly through the consumption of ready-to-eat foods such as cheese. Immunocompromised persons, the elderly, and pregnant women and their fetuses or newborns are at the highest risk for the infection. We examined the effects

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service