Skip to Content
Merck
All Photos(1)

Key Documents

P6534

Sigma-Aldrich

Phospholipase A2 from porcine pancreas

ammonium sulfate suspension, ≥600 units/mg protein

Synonym(s):

Lecithinase A, PLA2, Phosphatidylcholine 2-acylhydrolase

Sign Into View Organizational & Contract Pricing


About This Item

CAS Number:
Enzyme Commission number:
EC Number:
MDL number:
UNSPSC Code:
12352204
NACRES:
NA.32

form

ammonium sulfate suspension

specific activity

≥600 units/mg protein

UniProt accession no.

storage temp.

2-8°C

Gene Information

Looking for similar products? Visit Product Comparison Guide

General description

Phospholipase A2 is a small disulfide-rich protein having 124 residues. It is a calcium-dependent enzyme.

Application

Phospholipase A2 has been used in phospholipase assay and to determine rat renal proximal tubular segments (PTS) viability during oxygenation and hypoxia-reoxygenation.

Biochem/physiol Actions

Hydrolyzes the β-ester bond of zwitterionic glycerophospholipids. Preferred substrates are phosphatidylcholine, phosphatidylethanolamine, and their plasmalogen analogues. Phosphatidylinositol and phosphatidylserine are also hydrolyzed. Aggressively attacks phospholipids in membranes of intact cells.
It has a high catalytic activity on aggregated substrates compared to monomeric substrates.

Unit Definition

One unit will hydrolyze 1.0 μmole of soybean L-α-phosphatidylcholine to L-α-lysophosphatidylcholine and a fatty acid per min at pH 8.0 at 37 °C.

Physical form

Suspension in 3.2 M (NH4)2SO4 solution, pH 5.5

Analysis Note

Protein determined by biuret.

Storage Class Code

11 - Combustible Solids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

A A Grippo et al.
Journal of reproduction and fertility, 102(1), 87-93 (1994-09-01)
Cholesterol and phospholipid concentrations and phospholipase activity were measured in fluid from cannulae collected from the bovine oviductal isthmus and ampulla at different stages of the oestrous cycle. The cholesterol concentration and cholesterol normalized by protein were significantly (P =
B van den Berg et al.
Journal of biomolecular NMR, 5(2), 110-121 (1995-02-01)
The three-dimensional structure of porcine pancreatic PLA2 (PLA2), present in a 40 kDa ternary complex with micelles and a competitive inhibitor, has been determined using multidimensional heteronuclear NMR spectroscopy. The structure of the protein (124 residues) is based on 1854
Elena Venuti et al.
Journal of cystic fibrosis : official journal of the European Cystic Fibrosis Society, 16(6), 763-770 (2017-07-26)
Bile salt stimulated lipase (BSSL; Enzyme Commission (EC) number 3.1.1.13) has been a candidate triglyceridase for improving enzyme therapy for pancreatic insufficiency; however, its efficacy is near absent. We hypothesise that similarly to pancreatic lipase, BSSL is inhibited by phospholipids
Knut Kölbel et al.
Biophysical chemistry, 206, 12-21 (2015-06-29)
Porcine pancreatic phospholipase A2, a small and disulfide rich protein, is extremely resistant against chemically or thermally induced unfolding. Despite this marked resistance, the protein displays broad unfolding transitions resulting in comparatively low apparent thermodynamic stability. Broad unfolding transitions may
R A Zager et al.
Proceedings of the National Academy of Sciences of the United States of America, 90(17), 8297-8301 (1993-09-01)
During hypoxic or ischemic renal tubular injury, phospholipase A2 (PLA2) induces membrane deacylation, causing fatty acid accumulation and phospholipid breakdown. Because these changes can compromise cellular integrity, PLA2 activity has been widely proposed as a critical mediator of hypoxic renal

Articles

Instructions for working with enzymes supplied as ammonium sulfate suspensions

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service