Skip to Content
Merck
All Photos(2)

Key Documents

P6487

Sigma-Aldrich

Anti-Purinergic Receptor P2Y1 antibody produced in rabbit

affinity isolated antibody, lyophilized powder

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
NACRES:
NA.41

biological source

rabbit

Quality Level

conjugate

unconjugated

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

form

lyophilized powder

species reactivity

human, rat

technique(s)

immunohistochemistry: suitable
western blot: suitable

UniProt accession no.

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

human ... P2RY1(5028)
rat ... P2ry1(25265)

General description

Purinergic Receptor P2Y1 is a member of G protein-coupled receptors. The P2Y1 gene is mapped to human chromosome 3q25.2. It is present in the heart, blood vessels, prostate, testis, ovary, and neural tissue.

Immunogen

synthetic peptide corresponding to residues 242-258 of rat or human P2Y1.

Application

Anti-Purinergic Receptor P2Y1 antibody produced in rabbit has been used in western blotting1:200 and immunohistochemistry at a dilution of 1:50.

Biochem/physiol Actions

Purinergic Receptor P2Y1 acts as a receptor for extracellular ATP and ADP. It is essential for ADP-induced platelet activation, aggregation, and thrombosis. P2Y1  also participates in endothelial cell-based nucleotide-induced relaxation and in calcium mobilization. It serves as a potential target for the generation of new antiplatelet compounds and anti-inflammatory strategies.

Physical form

Lyophilized from phosphate buffered saline, pH 7.4, 1% bovine serum albumin and 0.05% sodium azide.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Hazard Statements

Precautionary Statements

Hazard Classifications

Aquatic Chronic 3

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

J Simon et al.
Acta biologica Hungarica, 54(2), 191-201 (2003-10-11)
Since the first identification of P2Y receptor sequences in 1993, it has quickly become apparent that this family of the G-protein coupled receptors is very diverse. Members of this receptor family are activated extra-cellularly by a wide variety of adenosine
Shara Khan et al.
Diabetologia, 57(12), 2535-2545 (2014-09-12)
There is evidence that ATP acts as an autocrine signal in beta cells but the receptors and pathways involved are incompletely understood. Here we investigate the receptor subtype(s) and mechanism(s) mediating the effects of ATP on human beta cells. We
Milka Koupenova et al.
Frontiers in pharmacology, 9, 37-37 (2018-02-15)
Platelets are small anucleated cells present only in mammals. Platelets mediate intravascular hemostatic balance, prevent interstitial bleeding, and have a major role in thrombosis. Activation of platelet purinergic receptors is instrumental in initiation of hemostasis and formation of the hemostatic
Anna Wilkaniec et al.
Purinergic signalling, 13(3), 347-361 (2017-05-19)
Abnormalities of alpha-synuclein (ASN), the main component of protein deposits (Lewy bodies), were observed in Parkinson's disease (PD), dementia with Lewy bodies, Alzheimer's disease, and other neurodegenerative disorders. These alterations include increase in the levels of soluble ASN oligomers in
Béatrice Hechler et al.
Purinergic signalling, 7(3), 293-303 (2011-07-28)
Following vessel wall injury, platelets adhere to the exposed subendothelium, become activated and release mediators such as TXA(2) and nucleotides stored at very high concentration in the so-called dense granules. Released nucleotides and other soluble agents act in a positive

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service