Skip to Content
Merck
All Photos(3)

Key Documents

P6402

Sigma-Aldrich

Perphenazine

Synonym(s):

4-[3-(2-Chloro-10H-phenothiazin-10-yl)propyl]-1-piperazineethanol

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C21H26ClN3OS
CAS Number:
Molecular Weight:
403.97
EC Number:
MDL number:
UNSPSC Code:
12352200
PubChem Substance ID:
NACRES:
NA.77

form

powder

originator

Schering Plough

storage temp.

2-8°C

SMILES string

ClC(C=C1)=CC2=C1SC3=CC=CC=C3N2CCCN4CCN(CCO)CC4

InChI

1S/C21H26ClN3OS/c22-17-6-7-21-19(16-17)25(18-4-1-2-5-20(18)27-21)9-3-8-23-10-12-24(13-11-23)14-15-26/h1-2,4-7,16,26H,3,8-15H2

InChI key

RGCVKNLCSQQDEP-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

Perphenazine is an antipsychotic drug used in treating schizophrenia. It modulates antioxidant levels and helps in the management of oxidative injury in schizophrenia. However, it is less effective than the clozapine drug group in improving response to oxidative injury.

Biochem/physiol Actions

D2 dopamine receptor antagonist; α-adrenergic receptor antagonist and σ-receptor agonist; phenothiazine antipsychotic. Inhibits glutamate dehydrogenase in vitro.

Features and Benefits

This compound was developed by Schering Plough. To browse the list of other pharma-developed compounds and Approved Drugs/Drug Candidates, click here.

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Acute Tox. 4 Oral - Skin Sens. 1

Storage Class Code

11 - Combustible Solids

WGK

WGK 3


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

J Nordenberg et al.
Biochemical pharmacology, 58(8), 1229-1236 (1999-09-16)
Some of the psychotropic agents widely used for the amelioration of anxiety, depression, and psychosis also show an effect at the cellular proliferation level. Surprisingly little research, however, has been directed to the antitumoral potential of these drugs, alone or
Donald E Addington et al.
The Journal of clinical psychiatry, 72(1), 75-80 (2010-09-28)
According to the American Psychiatric Association Clinical Practice Guidelines for schizophrenia, second-generation antipsychotics may be specifically indicated for the treatment of depression in schizophrenia. We examined the impact of these medications on symptoms of depression using the data from the
Angela M Koranek et al.
Schizophrenia research, 137(1-3), 137-140 (2012-03-01)
Results from the Clinical Antipsychotic Trials of Intervention Effectiveness (CATIE) indicate that, with the exception of olanzapine, no substantial overall differences were identified between second generation antipsychotics (SGAs) and the first generation antipsychotic (FGA) perphenazine. This study evaluated the effect
Elina Turunen et al.
The Journal of pharmacy and pharmacology, 63(1), 19-25 (2010-12-16)
The sublingual administration route as well as solid dispersion formation with macrogol 8000 and complexation with β-cyclodextrin (β-CyD) were investigated as ways for improving the absorption of perphenazine, a poorly water-soluble drug subjected to substantial first-pass metabolism. The absorption of
Alterations in oxidative stress markers and its correlation with clinical findings in schizophrenic patients consuming perphenazine, clozapine and risperidone
Hendouei N, et al.
Biomedicine and Pharmacotherapy, 103, 965-972 (2018)

Articles

Sigma article discusses tumor cell metabolic pathways, focusing on aerobic glycolysis and mitochondrial activity.

Sigma article discusses tumor cell metabolic pathways, focusing on aerobic glycolysis and mitochondrial activity.

Sigma article discusses tumor cell metabolic pathways, focusing on aerobic glycolysis and mitochondrial activity.

Sigma article discusses tumor cell metabolic pathways, focusing on aerobic glycolysis and mitochondrial activity.

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service