Skip to Content
Merck
All Photos(1)

Key Documents

P6244

Sigma-Aldrich

Protein Tyrosine Phosphatase 1B human

recombinant, expressed in E. coli

Synonym(s):

PTP-1B

Sign Into View Organizational & Contract Pricing


About This Item

MDL number:
UNSPSC Code:
12352204
NACRES:
NA.54

recombinant

expressed in E. coli

Quality Level

form

solution

specific activity

≥20 units/mg protein

mol wt

37.4 kDa

concentration

≥0.5 mg/mL

UniProt accession no.

relevant disease(s)

diabetes; obesity; cancer

shipped in

dry ice

storage temp.

−70°C

Gene Information

human ... PTPN1(5770)

Looking for similar products? Visit Product Comparison Guide

Related Categories

General description

Protein tyrosine phosphatase 1B human (PTP1B) is encoded by the gene PTPN1. In human chromosome, the gene PTPN1 is localized on 20q13.13. The protein has 37 kDa catalytic domain and the C-terminal hydrophobic residues targets the protein to the cytoplasmic side of the endoplasmic reticulum membrane.

Application

Protein Tyrosine Phosphatase 1B (PTP1B) human has been used in treating cell lysate prior to immunoprecipitation and protein pull-down studies. It has also been used in PTP1B activity assay and also for the evaluation of inhibitors like betulinic acid (BA) and sodium orthovanadate (Na3VO4), and 6-chloro-3-formyl-7-methylchromone (CFM) on PTP1B.
Protein tyrosine phosphatase 1B (PTP1B) is a pharmaceutical target used to study the treatment of various diseases such as obesity, cancer and type II diabetes mellitus.
The inhibitory effect of 8-(4-chlorophenylthio)-cAMP and 8-(4-chlorophenylthio)-cGMP on protein tyrosine phosphatase-1B has been assayed.

Biochem/physiol Actions

PTP-1B is a recombinant, non-transmembrane protein expressed in E. coli containing amino acid residues 1-322 of human PTP-1B. It has a molecular mass of 37.4 kDa. It is an abundant intracellular enzyme that is thought to act as a negative regulator of certain signaling pathways. It dephosphorylates tyrosine-phosphorylated proteins and peptides, and is usually localized in the cytostolic domain of the ER.
PTP1B dephosphorylates c-Src in several human breast cancer cell lines has a regulatory role for PTP1B in the control of c-Src kinase activity. PTP1B is regulates the cardiovascular effects of leptin. PTP1B controls the JAK/STAT signaling pathway by dephosphorylation of JAK2.
Polymorphism in the gene PTPN1 is associated with type 2 diabetes and obesity.

Unit Definition

One unit will hydrolyze 1.0 μmole of a phosphopeptide, EGFR fragment 988-998, per minute at pH 7.2 at 30 °C

Physical form

Supplied as a solution in 50 mM HEPES (pH 7.2), 1 mM EDTA, 5 mM DTT, and 0.05% NP-40

Storage Class Code

12 - Non Combustible Liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

p66Shc couples mechanical signals to RhoA through FAK-dependent recruitment of p115-RhoGEF and GEF-H1
Wu RF, et al.
Molecular and Cellular Biology, MCB-00194 (2016)
Analysis of common PTPN1 gene variants in type 2 diabetes, obesity and associated phenotypes in the French population
Cheyssac C, et al.
BMC Medical Genetics, 7(1), 44-44 (2006)
A colorimetric biosensor based on guanidinium recognition for the assay of protein tyrosine phosphatase 1B and its inhibitors
Lv J, et al.
New. J. Chem., 41(23), 14414-14419 (2017)
Adsorption between TC-stabilized AuNPs and the phosphate group: application of the PTP1B activity assay
Lv J, et al.
Analyst, 140(23), 8017-8022 (2015)
p66Shc couples mechanical signals to RhoA through FAK-dependent recruitment of p115-RhoGEF and GEF-H1
Wu RF, et al.
Molecular and cellular biology, MCB-00194 (2016)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service