Skip to Content
Merck
All Photos(3)

Key Documents

P4516

Sigma-Aldrich

Pullulan

suitable for substrate for pullulanase

Sign Into View Organizational & Contract Pricing


About This Item

CAS Number:
EC Number:
MDL number:
UNSPSC Code:
12352201
NACRES:
NA.25

form

powder or crystals

color

white

solubility

water: 50 mg/mL, colorless to very faintly yellow

suitability

suitable for substrate for pullulanase

storage temp.

2-8°C

Looking for similar products? Visit Product Comparison Guide

General description

Pullulan is a natural polysaccharide.

Application

Pullulan has been used in a study to assess bone tissue engineering. It has also been used in a study that investigated anti-fog applications.

Other Notes

To gain a comprehensive understanding of our extensive range of Polysaccharides for your research, we encourage you to visit our Carbohydrates Category page.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Laura Introzzi et al.
ACS applied materials & interfaces, 4(7), 3692-3700 (2012-07-05)
A new antifog coating made of pullulan is described in this work. The antifog properties are discussed in terms of wettability, surface chemistry/morphology, and by quantitative assessment of the optical properties (haze and transparency) before and after fog formation. The
Jean Christophe Fricain et al.
Biomaterials, 34(12), 2947-2959 (2013-02-05)
Research in bone tissue engineering is focused on the development of alternatives to allogenic and autologous bone grafts that can stimulate bone healing. Here, we present scaffolds composed of the natural hydrophilic polysaccharides pullulan and dextran, supplemented or not with
Masako Fujioka-Kobayashi et al.
Biomaterials, 33(30), 7613-7620 (2012-07-18)
To create a drug delivery system that allows the controlled release of proteins, such as growth factors, over a long-term period, cholesteryl group- and acryloyl group-bearing pullulan (CHPOA) nanogels were aggregated to form fast-degradable hydrogels (CHPOA/hydrogels) by cross-linking with thiol-bearing
Ricardo J B Pinto et al.
Colloids and surfaces. B, Biointerfaces, 103, 143-148 (2012-12-04)
Silver has been mainly investigated as an antibacterial agent and less as a fungicide in which concerns antimicrobial properties. In this research, the antifungal activity of composite films of pullulan and Ag nanoparticles (NP) against Aspergillus niger was evaluated using
Jeong-Hun Kang et al.
Journal of controlled release : official journal of the Controlled Release Society, 163(2), 119-124 (2012-09-15)
Apolipoprotein B (Apo B) is a key amphipathic glycoprotein compound in the metabolism of plasma lipoproteins (mainly very low-density lipoprotein (VLDL) and LDL). Inhibition of Apo B synthesis by short interfering RNA (siRNA) targeting Apo B (Apo B siRNA) is

Related Content

Dextrans are polysaccharides with molecular weights ≥1,000 Dalton, featuring a linear backbone of α-linked d-glucopyranosyl repeating units.

Dextrans are polysaccharides with molecular weights ≥1,000 Dalton, featuring a linear backbone of α-linked d-glucopyranosyl repeating units.

Dextrans are polysaccharides with molecular weights ≥1,000 Dalton, featuring a linear backbone of α-linked d-glucopyranosyl repeating units.

Dextrans are polysaccharides with molecular weights ≥1,000 Dalton, featuring a linear backbone of α-linked d-glucopyranosyl repeating units.

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service