Skip to Content
Merck
All Photos(2)

Key Documents

P4413

Sigma-Aldrich

N,N,N′,N′-Tetrakis(2-pyridylmethyl)ethylenediamine

≥98% (TLC), powder, membrane-permeable zinc chelator

Synonym(s):

TPEN

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C26H28N6
CAS Number:
Molecular Weight:
424.54
Beilstein:
4211982
MDL number:
UNSPSC Code:
12352116
PubChem Substance ID:
NACRES:
NA.77

Product Name

N,N,N′,N′-Tetrakis(2-pyridylmethyl)ethylenediamine,

form

powder

SMILES string

C(CN(Cc1ccccn1)Cc2ccccn2)N(Cc3ccccn3)Cc4ccccn4

InChI

1S/C26H28N6/c1-5-13-27-23(9-1)19-31(20-24-10-2-6-14-28-24)17-18-32(21-25-11-3-7-15-29-25)22-26-12-4-8-16-30-26/h1-16H,17-22H2

InChI key

CVRXLMUYFMERMJ-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Application

N,N,N′,N′-Tetrakis(2-pyridylmethyl)ethylenediamine has been used:
  • in the treatment of human embryonic kidney (HEK293T) cells for assessing its effect on the Jembrana disease virus (JDV) viral infectivity factor (Vif) functionality
  • in P19 embryonic carcinoma cells for chelation of zinc ions released during apoptosis
  • to evaluate its effect on lentiviral Vpx infection on human monocytic THP-1 cells

Biochem/physiol Actions

N,N,N′,N′-Tetrakis(2-pyridylmethyl)ethylenediamine (TPEN) elicits pro-apoptotic effect in human acute promyelocytic NB4 cells by inhibiting nitric oxide signalling and Zn2+ signaling. In microglia pretreated with zinc, TPEN inhibits the interleukin 6 secretion and nitric oxide synthase (iNOS) mRNA expression. It induces damage on DNA structure by activating ATM (ataxia telangiectasia mutated) and ATR (ATM-Rad3-related) signaling in human colon cancer cells.
TPEN is a membrane-permeable zinc chelator; decreased the intracellular level of zinc and induced apoptosis (i.e., cell shrinkage and formation of apoptotic bodies with DNA fragmentation and formation of a typical DNA ladder pattern).

Features and Benefits

This compound is a featured product for Apoptosis research. Click here to discover more featured Apoptosis products. Learn more about bioactive small molecules for other areas of research at sigma.com/discover-bsm.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Inhibition of Vpx-mediated SAMHD1 and Vpr-mediated HLTF degradation by selective disruption of viral CRL4 (DCAF1) E3 ubiquitin ligase assembly
Wang H, et al.
Journal of Virology, JVI-00225 (2017)
Xin Li et al.
Frontiers in molecular neuroscience, 10, 414-414 (2018-01-10)
Transient receptor potential melastatin-related 2 (TRPM2) channel, a molecular sensor for reactive oxygen species (ROS), plays an important role in cognitive dysfunction associated with post-ischemia brain damage thought to result from ROS-induced TRPM2-dependent neuronal death during reperfusion. Emerging evidence further
Nicholas Dietrich et al.
Nucleic acids research, 45(20), 11658-11672 (2017-10-05)
The essential element zinc plays critical roles in biology. High zinc homeostasis mechanisms are beginning to be defined in animals, but low zinc homeostasis is poorly characterized. We investigated low zinc homeostasis in Caenorhabditis elegans because the genome encodes 14
Synthesis of a highly Zn 2+-selective cyanine-based probe and its use for tracing endogenous zinc ions in cells and organisms
Guo Z, et al.
Nature Protocols, 9(6), 1245-1245 (2014)
Elodie Mattio et al.
Talanta, 183, 201-208 (2018-03-24)
In recent years, the development of 3D printing in flow analysis has allowed the creation of new systems with various applications. Up to now, 3D printing was mainly used for the manufacture of small units such as flow detection cells

Articles

Cell cycle phases (G1, S, G2, M) regulate cell growth, DNA replication, and division in proliferating cells.

Cell cycle phases (G1, S, G2, M) regulate cell growth, DNA replication, and division in proliferating cells.

Cell cycle phases (G1, S, G2, M) regulate cell growth, DNA replication, and division in proliferating cells.

Cell cycle phases (G1, S, G2, M) regulate cell growth, DNA replication, and division in proliferating cells.

See All

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service