Skip to Content
Merck
All Photos(1)

Key Documents

P4069

Sigma-Aldrich

Phosphatase, Alkaline from Escherichia coli

buffered aqueous glycerol solution, 20-50 units/mg protein (in glycine buffer)

Synonym(s):

Orthophosphoric-monoester phosphohydrolase (alkaline optimum)

Sign Into View Organizational & Contract Pricing


About This Item

CAS Number:
Enzyme Commission number:
EC Number:
MDL number:
UNSPSC Code:
12352204
NACRES:
NA.54
Pricing and availability is not currently available.

biological source

Escherichia coli

Quality Level

form

buffered aqueous glycerol solution

specific activity

20-50 units/mg protein (in glycine buffer)

foreign activity

DNase, RNase, none detected

storage temp.

−20°C

Looking for similar products? Visit Product Comparison Guide

Application

Alkaline phosphatase is used for conjugation to antibodies and other proteins for ELISA, Western blotting, and histochemical detection. It may be used for protein labeling when high sensitivity is required. Product P4069 is provided as a buffered aqueous glycerol solution.
Phosphatase, Alkaline from Escherichia coli has been used:
  • in fluorometric assay to determine its enzyme activity[1]
  • in the dephosphorylating enzyme cocktail for dephosphorylation of peptides[2]
  • in treating the nerve and HL60 cell lysate for β-hydroxy β-methylglutaryl-Coenzyme A (HMG-CoA) reductase activity[3]

Biochem/physiol Actions

Alkaline phosphatase is a model enzyme for understanding phosphomonoesterase. It is used in various biochemical methods and enzyme linked immunosorbent assay (ELISA).[1]
Alkaline phosphatase, from Escherichia coli, is a dimeric, non-glycosylated protein which mainly reside in the periplasmic space. Three known isoforms exist. The enzyme requires zinc, and is activated by magnesium. E. coli akaline phosphatase has a broad specificity for phosphate esters.

Caution

Alkaline phosphatase in 5 mM EDTA, pH 8, will be irreversibly heat inactivated at 75°C for 10 minutes.

Unit Definition

One unit will hydrolyze 1.0 μmole of p-nitrophenyl phosphate per min at pH 10.4 at 37 °C.

Physical form

Solution in 5 mM Tris-HCl, pH 7.4, 0.5 mM MgCl2, 0.5 mM ZnCl2, 50% (w/v) glycerol

Analysis Note

Protein determined by biuret.

Pictograms

Health hazard

Signal Word

Danger

Hazard Statements

Precautionary Statements

Hazard Classifications

Resp. Sens. 1

Storage Class Code

10 - Combustible liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

The prion protein and cellular cholesterol homeostasis
Diomede L, et al.
Neurobiology of Lipids, 1(2), 8-14 (2002)
Sensitive fluorogenic substrate for alkaline phosphatase
Levine MN and Raines RT
Analytical Biochemistry, 418(2), 247-252 (2011)
Phospho-iTRAQ data article: Assessing isobaric labels for the large-scale study of phosphopeptide stoichiometry
Glibert P, et al.
Data in Brief, 4(2), 60-65 (2015)
Lionel M Igaz et al.
The American journal of pathology, 173(1), 182-194 (2008-06-07)
TAR DNA-binding protein (TDP-43) has been recently described as a major pathological protein in both frontotemporal dementia with ubiquitin-positive inclusions (FTLD-U) and amyotrophic lateral sclerosis. However, little is known about the relative abundance and distribution of different pathological TDP-43 species
A D Bakker et al.
Journal of dental research, 93(4), 394-399 (2014-02-05)
Mechanosensitive osteocytes regulate bone mass in adults. Interleukin 6 (IL-6), such as present during orthodontic tooth movement, also strongly affects bone mass, but little is known about the effect of IL-6 on osteocyte function. Therefore we aimed to determine in

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service