Skip to Content
Merck
All Photos(1)

Key Documents

P3237

Sigma-Aldrich

Anti-phospho-PAK1 (pThr212) antibody, Mouse monoclonal

clone PK-18, purified from hybridoma cell culture

Sign Into View Organizational & Contract Pricing


About This Item

MDL number:
UNSPSC Code:
12352203
NACRES:
NA.44

biological source

mouse

Quality Level

conjugate

unconjugated

antibody form

purified from hybridoma cell culture

antibody product type

primary antibodies

clone

PK-18, monoclonal

form

buffered aqueous solution

mol wt

antigen 68 kDa

species reactivity

human, mouse, rat

concentration

~2 mg/mL

technique(s)

immunocytochemistry: suitable
indirect ELISA: suitable
microarray: suitable
western blot: 0.2-0.4 μg/mL using a whole extract of transfected and activated COS-7 (monkey kidney) cells expressing phosphorylated PAK1

isotype

IgG1

UniProt accession no.

shipped in

dry ice

storage temp.

−20°C

target post-translational modification

phosphorylation (pThr212)

Gene Information

human ... PAK1(5058)
mouse ... Pak1(18479)
rat ... Pak1(29431)

General description

Monoclonal Anti-Phospho-PAK1 (pThr212) (mouse IgG1 isotype) is derived from the PK-18 hybridoma produced by the fusion of mouse myeloma cells and splenocytes from BALB/c mice immunized with a synthetic peptide corresponding to amino acids of human PAK1,1 conjugated to keyhole limpet hemocyanin (KLH). PAK1 (p21-activated kinase 1) is homologous to Sacharomyces cerevisiae kinase Ste20. PAK1 (68 kDa),with a C-terminal serine/threonine kinase domain and a N-terminal CRIB (Cdc42/Rac interactive binding) domain. Thr212 localized in flexible region is of PAK1 domain.
p21-activated kinases are a family of highly conserved serine/threonine protein kinases that are activated by small GTPases of Cdc42 and Rac families. There are 2 groups of PAKs in mammals, group A and B. The group A members are three isoforms, PAK1, PAK2, and PAK3 (also known as α-PAK, γ-PAK, and β-PAK, respectively). PAKs are actively involved in regulation of MAPK pathways by phosphorylation of Raf-1 and MEK-1. They also regulate cytoskeletal reorganization by interaction with GTPases. Autophosphorylation of threonine 423 (threonine 402 for PAK2 and threonine 421 for PAK3), catalyzed by Cdc42, is required for activation of PAK. In the neurons, the reorganization of cytoskeleton during the neurite outgrowth is modulated by phosphorylation of PAK1 at Thr212 that regulates a specific signalling pathway
Monoclonal Anti-Phospho-PAK1 (pThr212) reacts specifically with human PAK1 phosphorylated at Thr212 (68 kDa), and does not detect the unphosphorylated epitope.

Immunogen

synthetic peptide corresponding to amino acids 203-217 (pThr212) of human PAK1.

Application

Anti-phospho-PAK1 (pThr212) antibody, Mouse monoclonal has been used in:
  • enzyme linked immunosorbent assay (ELISA)
  • immunoblotting
  • immunohistochemistry


Anti-phospho-PAK1 (pThr212) may be used for immunoblotting at a working concentration of 0.2-0.4 μg/mL. The antibody is also suitable for indirect ELISA, immunocytochemistry and protein microarray.

Biochem/physiol Actions

p21-activated kinase 1 (PAK1) also regulates cytoskeletal dynamics by decreasing MLCK (myosin light chain kinase) activity and MLC phosphorylation. PAK1 is reported to be a crucial signaling molecule, involved in the transcription factor nuclear factor κB (NFκB) activation by multiple stimuli. PAK1 regulates cell morphology and polarity in eukaryotic cells and activates downstream targets like JNK (c-Jun N-terminal protein kinase) and p38 mitogen-activated protein kinase (MAPK).. The phosphorylation of actin by PAK1 at thr212 helps in regulating growth cone dynamics.

Physical form

Solution in 0.01 M phosphate buffered saline, pH 7.4, containing 15 mM sodium azide.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

P21 activated kinases: structure, regulation, and functions
Rane CK and Minden A
Small Gtpases, 5(1), e28003-e28003 (2014)
Phosphorylation of Pak1 by the p35/Cdk5 kinase affects neuronal morphology
Rashid T, et al.
Test, 276(52), 49043-49052 (2001)
Activation of p21-activated kinase 1-nuclear factor kappaB signaling by Kaposi?s sarcoma-associated herpes virus G protein-coupled receptor during cellular transformation
Dadke D, et al.
Cancer Research, 63(24), 8837-8847 (2003)
Activating mutations in PAK1, encoding p21-activated kinase 1, cause a neurodevelopmental disorder
Harms FL, et al.
American Journal of Human Genetics, 103(4), 579-591 (2018)
Susanne Hipp et al.
Journal of cellular and molecular medicine, 13(9B), 3858-3867 (2009-07-17)
Reduced E-cadherin expression is associated with tumour progression of many carcinomas, including endometrial cancers. The transcription factor Snail is known as one of the most prominent transcriptional E-cadherin repressors; its regulation in cancer tissues, however, still remains unclear. Here, we

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service