Skip to Content
Merck
All Photos(1)

Key Documents

P2014

Sigma-Aldrich

Phosphorylase Kinase from rabbit muscle

lyophilized powder, ≥60 units/mg protein

Synonym(s):

ATP:phosphorylase-b phosphotransferase, Dephosphophosphorylase kinase

Sign Into View Organizational & Contract Pricing


About This Item

CAS Number:
Enzyme Commission number:
MDL number:
UNSPSC Code:
12352204
NACRES:
NA.54

form

lyophilized powder

specific activity

≥60 units/mg protein

composition

Protein, 20-40% biuret

foreign activity

ATPase ≤0.5%
phosphorylase a ≤1%
phosphorylase b ≤5%

storage temp.

−20°C

General description

Phosphorylase kinase contains four subunits each containing an α, β, γ, and sigma component. The sigma component binds 4 calcium molecules and is termed calmodulin while the γ unit acts as the catalytic subunit.

Application

Phosphorylase kinase from rabbit muscle has been used in a study to assess features of glycogen phosphorylase. It has also been used in a study to investigate the activation of different forms of muscle phosphorylase kinase by actin.

Unit Definition

One unit will form 1.0 μmolar unit of phosphorylase a from phosphorylase b per min at pH 7.7 at 30°C in the presence of ATP.

Physical form

Lyophilized powder containing (NH4)2SO4, sucrose, β-glycerophosphate and dithioerythritol

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

S Camus et al.
Oncogene, 31(39), 4333-4342 (2011-12-20)
Angiogenesis is essential for development and tumor progression. With the aim of identifying new compound inhibitors of the angiogenesis process, we used an established enhanced green fluorescent protein-transgenic zebrafish line to develop an automated assay that enables high-throughput screening of
K F Chan et al.
The Journal of biological chemistry, 257(10), 5956-5961 (1982-05-25)
Aspects of the molecular interaction and subunit structure of rabbit skeletal muscle phosphorylase kinase, (alpha beta gamma delta)4, were investigated. Exogenous addition of the delta subunit (calmodulin) stimulated the activities of nonactivated phosphorylase kinase and the alpha gamma delta complex
G Kamp
Biological chemistry Hoppe-Seyler, 367(2), 109-117 (1986-02-01)
The activities of glycogen phosphorylases a and b from the body wall musculature of the marine worm Arenicola marina (Annelida, Polychaeta) were determined after various periods of anoxia. Already under normoxic conditions one third of the total activity was produced
Simona Fermani et al.
The Journal of biological chemistry, 287(25), 21372-21383 (2012-04-20)
Carbon assimilation in plants is regulated by the reduction of specific protein disulfides by light and their re-oxidation in the dark. The redox switch CP12 is an intrinsically disordered protein that can form two disulfide bridges. In the dark oxidized
Kathryn A Skelding et al.
Advances in experimental medicine and biology, 740, 703-730 (2012-03-29)
Calcium/calmodulin-stimulated protein kinases can be classified as one of two types - restricted or multifunctional. This family of kinases contains several structural similarities: all possess a calmodulin binding motif and an autoinhibitory region. In addition, all of the calcium/calmodulin-stimulated protein

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service