Skip to Content
Merck
All Photos(1)

Key Documents

P1761

Sigma-Aldrich

trans-2-Phenylcyclopropylamine hemisulfate salt

Synonym(s):

Tranylcypromine

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
C9H11N · 1/2H2SO4
CAS Number:
Molecular Weight:
182.23
EC Number:
MDL number:
UNSPSC Code:
12352116
PubChem Substance ID:
NACRES:
NA.77

Assay

≥98% (perchloric acid titration)

Quality Level

form

powder

solubility

water: 0.0333 g/mL, clear to slightly hazy, colorless to faintly yellow

storage temp.

2-8°C

SMILES string

OS(O)(=O)=O.N[C@H]1C[C@@H]1c2ccccc2.N[C@H]3C[C@@H]3c4ccccc4

InChI

1S/2C9H11N.H2O4S/c2*10-9-6-8(9)7-4-2-1-3-5-7;1-5(2,3)4/h2*1-5,8-9H,6,10H2;(H2,1,2,3,4)/t2*8-,9+;/m11./s1

InChI key

BKPRVQDIOGQWTG-KAVFMPKWSA-N

Looking for similar products? Visit Product Comparison Guide

Biochem/physiol Actions

trans-2-Phenylcyclopropylamine acts as an inhibitor of lysine-specific demethylase 1 (LSD1). It exhibits anti-depressant and anxiolytic activity. Tranylcypromine shows therapeutic effects against mood and anxiety issues. It also exhibits anti-osteoclastogenic effects through monoamine oxidase-A (MAO-A). Tranylcypromine may play a role in bone remodeling in mice.
Non-selective MAO-A/B inhibitor.

Pictograms

Skull and crossbones

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 2 Oral - Acute Tox. 3 Dermal - Acute Tox. 3 Inhalation

Storage Class Code

6.1A - Combustible acute toxic Cat. 1 and 2 / very toxic hazardous materials

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Zhuochao Liu et al.
FASEB journal : official publication of the Federation of American Societies for Experimental Biology, 33(9), 9828-9841 (2019-07-11)
Identification of anti-osteoclastogenic agents is important for the treatment of bone loss diseases that feature excessive osteoclast (OC) activity and bone resorption. Tranylcypromine (TCP), an irreversible inhibitor of monoamine oxidase (MAO), has been used as an antidepressant and anxiolytic agent
Maojun Yang et al.
Biochemistry, 46(27), 8058-8065 (2007-06-16)
Histone modifications, such as acetylation and methylation, are important epigenetic marks that regulate diverse biological processes that use chromatin as the template, including transcription. Dysregulation of histone acetylation and methylation leads to the silencing of tumor suppressor genes and contributes
Elisa Maria Rieckhoff et al.
Current biology : CB, 30(24), 4973-4983 (2020-11-21)
Cellular organelles such as the mitotic spindle adjust their size to the dimensions of the cell. It is widely understood that spindle scaling is governed by regulation of microtubule polymerization. Here, we use quantitative microscopy in living zebrafish embryos and

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service