Skip to Content
Merck
All Photos(3)

Key Documents

P1162

Sigma-Aldrich

Palmitic acid N-hydroxysuccinimide ester

≥98% (TLC)

Synonym(s):

N-(Palmitoyloxy)succinimide, N-Succinimidyl palmitate, Palmitic acid N-succinimidyl ester

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C20H35NO4
CAS Number:
Molecular Weight:
353.50
Beilstein:
1547411
MDL number:
UNSPSC Code:
12352211
PubChem Substance ID:
NACRES:
NA.25

Quality Level

Assay

≥98% (TLC)

form

powder

lipid type

saturated FAs

storage temp.

−20°C

SMILES string

CCCCCCCCCCCCCCCC(=O)ON1C(=O)CCC1=O

InChI

1S/C20H35NO4/c1-2-3-4-5-6-7-8-9-10-11-12-13-14-15-20(24)25-21-18(22)16-17-19(21)23/h2-17H2,1H3

InChI key

OTNHQVHEZCBZQU-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Application

Palmitic acid N-hydroxysuccinimide ester has been used to couple a protein for fabrication of targeted delivery vehicles. It has also been used in the covalent attachment of palmitic acid to ovalbumin.
Useful in the synthesis of hexadecanoyl derivatives of amino- and thio-compounds such as amino acids, aminoacyl-tRNA, coenzyme A, thioglycolic acid, etc.

Biochem/physiol Actions

Palmitic acid N-hydroxysuccinimide ester is used in the covalent linking of protein to palmitic acid.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Slide 1 of 1

1 of 1

Preparation of immune stimulating complexes (ISCOMs) as adjuvants
Mowat AM and Reid G
Current Protocols in Immunology, 16(1), 2-11 (1995)
Adam Edelstein et al.
Shock (Augusta, Ga.), 36(5), 451-457 (2011-09-23)
Translocation of bacteria and other luminal factors from the intestine following surgical injury can be a major driver of critical illness. Bile acids have been shown to play a key role in the loss of intestinal epithelial barrier function during
Cell-specific siRNA delivery by peptides and antibodies
Test, 502, 91-122 (2012)
Cell-specific siRNA delivery by peptides and antibodies
Methods in Enzymology, 502, 91-122 (2012)
Ana Rute Neves et al.
Nanotechnology, 26(49), 495103-495103 (2015-11-18)
Nanotechnology can be an important tool to improve the permeability of some drugs for the blood-brain barrier. In this work we created a new system to enter the brain by functionalizing solid lipid nanoparticles with apolipoprotein E, aiming to enhance

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service