Skip to Content
Merck
All Photos(1)

Key Documents

P0032

Sigma-Aldrich

Pyr3

≥98% (HPLC)

Synonym(s):

Ethyl-1-(4-(2,3,3-trichloroacrylamide)phenyl)-5-(trifluoromethyl)-1H-pyrazole-4-carboxylate

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C16H11Cl3F3N3O3
Molecular Weight:
456.63
MDL number:
UNSPSC Code:
12352200
PubChem Substance ID:
NACRES:
NA.25

Quality Level

Assay

≥98% (HPLC)

form

powder

color

white to beige

solubility

DMSO: 20 mg/mL, clear

storage temp.

2-8°C

SMILES string

CCOC(=O)c1cnn(-c2ccc(NC(=O)\C(Cl)=C(\Cl)Cl)cc2)c1C(F)(F)F

InChI

1S/C16H11Cl3F3N3O3/c1-2-28-15(27)10-7-23-25(12(10)16(20,21)22)9-5-3-8(4-6-9)24-14(26)11(17)13(18)19/h3-7H,2H2,1H3,(H,24,26)

InChI key

RZHGONNSASQOAY-UHFFFAOYSA-N

Biochem/physiol Actions

Pyr3 is a pyrazole compound that potently and selectively antagonizes TRPC3. Pyr3 inhibits TRPC3 by binding to the extracellular side of the receptor, resulting in suppression of B cell activation and cardiac hypertrophy.
Pyr3 is a pyrazole compound that potently and selectively antagonizes TRPC3. Pyr3 inhibits TRPC3 by binding to the extracellular side of the receptor, resulting in suppression of B cell activation and cardiac hypertrophy.

Members of the canonical transient receptor potential (TRPC) channel family are ion channels which conduct Ca+2 and are activated by membrane receptor-mediated stimulation of phospholipase C (PLC) activity. However, TRPC3 and other family members are also activated by membrane-independent diacylglycerol (DAG). BTP1 and BTP2 are pyrazoles that block TRPC channels, but they are not specific for TRPC subtypes. This channel is implicated in various processes, including B cell receptor (BCR)-mediated Ca+2 oscillations, activation of nuclear factor of activated T cells (NFAT), and promotion of cardiac hypertrophy.

Storage Class Code

11 - Combustible Solids

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Yi-Fan Wu et al.
Cell & bioscience, 7, 28-28 (2017-05-27)
Chloroquine, a bitter tastant, inhibits Ca In this study, thapsigargin (TG), but not caffeine, induced sustained intracellular Ca These results indicate that chloroquine inhibits Ca
Maria Cristina D Picardo et al.
PLoS biology, 17(2), e2006094-e2006094 (2019-02-23)
Inspiratory breathing movements depend on pre-Bötzinger complex (preBötC) interneurons that express calcium (Ca2+)-activated nonselective cationic current (ICAN) to generate robust neural bursts. Hypothesized to be rhythmogenic, reducing ICAN is predicted to slow down or stop breathing; its contributions to motor
Cheng Cai et al.
Archives of toxicology, 93(4), 1009-1020 (2019-02-27)
Particulate matter (PM) is one of the most important environmental issues worldwide, which is associated with not only pulmonary but also cardiovascular diseases. However, the underlying biological mechanisms of PM related cardiovascular dysfunction remained poorly defined, especially mediated by the
Naoya Hirata et al.
International journal of molecular sciences, 23(4) (2022-02-27)
Triple-negative breast cancer (TNBC) is a highly aggressive cancer for which targeted therapeutic agents are limited. Growing evidence suggests that TNBC originates from breast cancer stem cells (BCSCs), and elucidation of the molecular mechanisms controlling BCSC proliferation will be crucial
Wenya Du et al.
Molecular therapy : the journal of the American Society of Gene Therapy, 30(4), 1754-1774 (2022-01-26)
Acute pancreatitis (AP) is a common digestive disease without specific treatment, and its pathogenesis features multiple deleterious amplification loops dependent on translation, triggered by cytosolic Ca2+ ([Ca2+]i) overload; however, the underlying mechanisms in Ca2+ overload of AP remains incompletely understood.

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service