O4003
Sodium octyl sulfate
≥95%
Synonym(s):
Octyl sodium sulfate, SOS, Sodium capryl sulfate, Octyl sulfate sodium salt
About This Item
Recommended Products
description
anionic
Quality Level
Assay
≥95%
form
powder
mol wt
232.27 g/mol
mp
195 °C (dec.) (lit.)
solubility
H2O: 20 mg/mL
SMILES string
[Na+].CCCCCCCCOS([O-])(=O)=O
InChI
1S/C8H18O4S.Na/c1-2-3-4-5-6-7-8-12-13(9,10)11;/h2-8H2,1H3,(H,9,10,11);/q;+1/p-1
InChI key
WFRKJMRGXGWHBM-UHFFFAOYSA-M
Looking for similar products? Visit Product Comparison Guide
General description
Application
Storage Class Code
11 - Combustible Solids
WGK
WGK 2
Personal Protective Equipment
Choose from one of the most recent versions:
Certificates of Analysis (COA)
Don't see the Right Version?
If you require a particular version, you can look up a specific certificate by the Lot or Batch number.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Customers Also Viewed
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service